Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | ₪1,163.00 |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | ₪2,093.00 |
About This Item
₪1,163.00
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CTACATTAATAGGGTTAGGAGGGTTAAAGGGTAAAGAGTCCTTTGAGCACATTTTCTGGCAAACCATAATAACTTGATAAATGCTAAGCATTGGAACGGGTACCAAGTCTCCAAGAAACAAGTATCTCAATGCAGTTTTAGAAATTTCCTGTTTTATTCTTTTCTGTGGCCCTGCTCACCCTCTGGGAATAATCATTTCAACTTTGCATTTGTGAAAAATAATACATACAATCTTAAGGTTACTTCAGACTATACTTTTTCATCGAGGAGTATTGTATTCAAAAGTGATCGATCTGCTGAATATATAAATTAAAATGGACACAAGCCTTCATTCATCAGACATTTGAGTGTCTGTCCTCTGAGTTCCCATAGAATCTGAATTACACCACTTTATTCACACTTACATTATGTGTTTACTTTTCTGAGTGTCCTATTAATCTTTATTCCTTAAAGGAAAGCCTGAGGGCATCTTGTTTAACAATGCATCTCTCTCTTCATCAATCAACGCTG
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... TMEM236(653567), FAM23A()
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | ||
|---|---|---|
| description targeting human TMEM236 | description targeting mouse Fam23a | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody |
| Gene Information human ... TMEM236(653567), FAM23A() | Gene Information mouse ... FAM23A(625286), Fam23a(625286) | Gene Information human ... TMEM236(653567) |
| product line MISSION® | product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies |
| esiRNA cDNA target sequence CTACATTAATAGGGTTAGGAGGGTTAAAGGGTAAAGAGTCCTTTGAGCACATTTTCTGGCAAACCATAATAACTTGATAAATGCTAAGCATTGGAACGGGTACCAAGTCTCCAAGAAACAAGTATCTCAATGCAGTTTTAGAAATTTCCTGTTTTATTCTTTTCTGTGGCCCTGCTCACCCTCTGGGAATAATCATTTCAACTTTGCATTTGTGAAAAATAATACATACAATCTTAAGGTTACTTCAGACTATACTTTTTCATCGAGGAGTATTGTATTCAAAAGTGATCGATCTGCTGAATATATAAATTAAAATGGACACAAGCCTTCATTCATCAGACATTTGAGTGTCTGTCCTCTGAGTTCCCATAGAATCTGAATTACACCACTTTATTCACACTTACATTATGTGTTTACTTTTCTGAGTGTCCTATTAATCTTTATTCCTTAAAGGAAAGCCTGAGGGCATCTTGTTTAACAATGCATCTCTCTCTTCATCAATCAACGCTG | esiRNA cDNA target sequence AGTAGCACAAACCTAACAATGAGCTACATCCCTTAGAAATACATTTCTTACTCTGTTGCAGTTACCGCTACTAAGTCATTAGCTACATAGAAAACTAGGGTCTTCAGCACACATCAGTTTGGCAAAGCTTCCTTACAGCTTCTTGGGCTGGTGCAGCTGGCAGTGCGTCAGTGTTGAGGATAATGGAGCCAAGGTACATTGTGATTCCTAGATAAAGCTTTGTAATGGACACCATAAACAATGTGTAAGAGTGTAACATAAACAGAGTTTGATGCCATCATAAAAAAAAAATCAAGTAGTTTGCACTTAGTATAGATGCCAAAGGTTTCTTACGTTGTACTAGGTCCTGAGTATCTTCATGTTACAATGATCATGCACACAGTCATGAACTCTTCATGTGCTCTGATGGCATAGATACTCATATCCAAGCAGAGATTCTTTGTTTATAAGAGAAACTTGTAAGAAGTTTATCTCTGCAGAGTAATGTGGAAGCAAATAGCTTATGAACAG | esiRNA cDNA target sequence - |
| Ensembl | human accession no. | Ensembl | human accession no. - | Ensembl | human accession no. - |
| form lyophilized powder | form lyophilized powder | form buffered aqueous glycerol solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. - |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

