Merck
  • The effects of Caulerpa microphysa enzyme-digested extracts on ACE-inhibitory activity and in vitro anti-tumour properties.
The effects of Caulerpa microphysa enzyme-digested extracts on ACE-inhibitory activity and in vitro anti-tumour properties.
Food chemistry (2013-02-28)
Hui-Chiu Lin, Su-Tze Chou, Mei-Yu Chuang, Tzu-Yen Liao, Wann-Sheng Tsai, Tsai-Hsin Chiu
ABSTRACT
The aim of this study was to determine the ACE inhibitory activity and its anti-cancer properties of Caulerpa microphysa extracts. C. microphysa samples were digested with Flavourzyme, Alcalase, and pepsin. The ACE inhibitory activity of enzyme-digested C. microphysa decreased in the order of digestion with pepsin>Flavourzyme>Alcalase; that is, pepsin-extracted samples had significantly higher activity than the other enzyme extractions. To test its anti-tumour effects in vitro C. microphysa pepsin-digested extracts were applied to BALB/c mice with transplanted myelomonocytic leukaemia (WEHI-3) and Human promyelocytic leukaemia (HL-60) cell lines. The growth of both cell lines was inhibited, and extracts induced DNA damage, evaluated with a comet assay. The data demonstrate that C. microphysa pepsin-digested extract had the ability to anti-tumour effects. Further application as a health food is worthy of investigation.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein