Merck
  • Relationship between protein digestibility and allergenicity: comparisons of pepsin and cathepsin.
Relationship between protein digestibility and allergenicity: comparisons of pepsin and cathepsin.
Toxicology (2013-04-30)
Emily S Foster, Ian Kimber, Rebecca J Dearman
ABSTRACT
An association between protein allergenicity and resistance to pepsin digestion in the gastrointestinal tract has been proposed. However, although widely accepted, such an association is inconsistent with known labile allergens and resistant nonallergens. Given the central role of antigen presenting cells, and in particular dendritic cells (DC), in the development of allergic responses, the stability of allergens to intracellular processing may be more relevant than resistance to extracellular pepsin digestion. We have characterised the expression by DC of cathepsins (proteolytic enzymes), and compared the proteolytic activity of the most highly expressed cathepsin with pepsin for a range of 9 allergens and 4 putative nonallergens. Cathepsin expression in bone marrow-derived DC (BM-DC) derived from BALB/c strain mice was characterised by flow cytometry; cathepsins D, E and S were identified, with cathepsin D being the most highly expressed. Digestion studies revealed that the majority of allergens (5/9) were pepsin resistant, whereas non-allergens (3/4) were labile. If the generation of pepsin-resistant fragments was considered as a feature of allergenicity, this increased to 7/9 allergens and 4/4 nonallergens. In contrast, most of the proteins examined were resistant to cathepsin digestion, with significant digestion recorded for only 2/9 allergens and 2/4 non-allergens. Chemical reduction (to mimic intracellular reducing conditions) increased the susceptibility of proteins to digestion by cathepsins, but did not improve discrimination between allergens and nonallergens on this basis. These data confirm that there is a general relationship between resistance to digestion with pepsin and allergenicity. The relationship is not absolute, but the information gained from this characteristic does provide useful information in a weight of evidence approach for allergenicity assessment. The most abundant cathepsin detected in antigen processing BM-DC, cathepsin D, is not an appropriate substitute for pepsin. The hypothesis that pepsin stability may be a surrogate for stability to digestion within DC may still hold true, but consideration of a single enzyme in this context is possibly an oversimplification.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, lyophilized powder, ≥3,200 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥400 units/mg protein
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, ≥250 units/mg solid
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, 1200-2400 U/mg
Sigma-Aldrich
Pepsin from porcine gastric mucosa, tested according to Ph. Eur.
Sigma-Aldrich
Pepsin from porcine gastric mucosa, powder, slightly beige, ≥500 U/mg