Merck
Osteogenic properties of a short BMP-2 chimera peptide.
Journal of peptide science : an official publication of the European Peptide Society (2015-08-22)
Lucia Falcigno, Gabriella D'Auria, Luisa Calvanese, Daniela Marasco, Roberta Iacobelli, Pasqualina L Scognamiglio, Paola Brun, Roberta Danesin, Matteo Pasqualin, Ignazio Castagliuolo, Monica Dettin
ABSTRACT
Bone morphogenetic proteins (BMPs) play a key role in bone and cartilage formation. For these properties, BMPs are employed in the field of tissue engineering to induce bone regeneration in damaged tissues. To overcome drawbacks due to the use of entire proteins, synthetic peptides derived from their parent BMPs have come out as promising molecules for biomaterial design. On the structural ground of the experimental BMP-2 receptor complexes reported in the literature, we designed three peptides, reproducing the BMP-2 region responsible for the binding to the type II receptor, ActRIIB. These peptides were characterized by NMR, and the structural features of the peptide-receptor binding interface were highlighted by docking experiments. Peptide-receptor binding affinities were analyzed by means of ELISA and surface plasmon resonance techniques. Furthermore, cellular assays were performed to assess their osteoinductive properties. A chimera peptide, obtained by combining the sequence portions 73-92 and 30-34 of BMP-2, shows the best affinity for ActRIIB in the series and represents a good starting point for the design of new compounds able to reproduce osteogenic properties of the parent BMP-2.
Product Description
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
1-Propanol, ≥99%, FG
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
1-Propanol, natural, ≥98%, FG
Sigma-Aldrich
1-Propanol, anhydrous, 99.7%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%