Skip to Content
Merck

GF95863821

Palladium

foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%

Synonym(s):

Palladium, PD000275

Sign In to View Organizational & Contract Pricing.

Select a Size

1 EA

¥247,000

¥247,000


Check Cart for Availability

Request a Bulk Order

About This Item

Linear Formula:
Pd
CAS Number:
Molecular Weight:
106.42
NACRES:
NA.23
PubChem Substance ID:
UNSPSC Code:
12141733
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Palladium, foil, 50x50mm, thickness 0.1mm, as rolled, 99.95%

InChI

1S/Pd

SMILES string

[Pd]

InChI key

KDLHZDBZIXYQEI-UHFFFAOYSA-N

assay

≥99.95%

form

foil

manufacturer/tradename

Goodfellow 958-638-21

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

mp

1554 °C (lit.)

density

12.02 g/cm3 (lit.)

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF93419792GF94882955GF31033112
assay

≥99.95%

assay

≥99.95%

assay

≥99.95%

assay

99.99%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 958-638-21

manufacturer/tradename

Goodfellow 934-197-92

manufacturer/tradename

Goodfellow 948-829-55

manufacturer/tradename

Goodfellow 310-331-12

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

resistivity

9.96 μΩ-cm, 20°C

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

bp

2970 °C (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

density

12.02 g/cm3 (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Storage Class

13 - Non Combustible Solids

wgk

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xiao-Feng Wu et al.
ChemSusChem, 6(2), 229-241 (2013-01-12)
Palladium-catalyzed coupling reactions have become a powerful tool for advanced organic synthesis. This type of reaction is of significant value for the preparation of pharmaceuticals, agrochemicals, as well as advanced materials. Both, academic as well as industrial laboratories continuously investigate
Qing-An Chen et al.
Chemical Society reviews, 42(2), 497-511 (2012-11-10)
The transition metal catalyzed asymmetric hydrogenation of unsaturated compounds arguably presents one of the most attractive methods for the synthesis of chiral compounds. Over the last few decades, Pd has gradually grown up as a new and popular metal catalyst
Pazhamalai Anbarasan et al.
Chemical Society reviews, 40(10), 5049-5067 (2011-04-30)
The palladium-catalyzed cyanation of Ar-X (X = I, Br, Cl, OTf, and H) allows for an efficient access towards benzonitriles. After its discovery in 1973 and following significant improvements in recent decades, this methodology has become nowadays the most popular
Palladium(II)-catalyzed alkene functionalization via nucleopalladation: stereochemical pathways and enantioselective catalytic applications.
Richard I McDonald et al.
Chemical reviews, 111(4), 2981-3019 (2011-03-25)
Stephan Enthaler et al.
Chemical Society reviews, 40(10), 4912-4924 (2011-06-07)
The formation of oxygen-carbon bonds is one of the fundamental transformations in organic synthesis. In this regard the application of palladium-based catalysts has been extensively studied during recent years. Nowadays it is an established methodology and the success has been

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service