Skip to Content
Merck

Skip To

E1908

Anti-ELMO (C-terminal) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CED12, Anti-ELMO1, Anti-Engulfment and cell motility 1

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352203


biological source

rabbit

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~75 kDa

species reactivity

rat, mouse, human

technique(s)

immunoprecipitation (IP): 1-2 μL using whole extract of mouse A-20 cells., western blot: 1:1,000-1:2,000 using whole extract of rat brain, western blot: 1:2,000-1:3,000 using whole extract of human Jurkat cells.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ELMO1(9844)
mouse ... Elmo1(140580)
rat ... Elmo1(361251)

General description

ELMO1 (engulfment and cell motility 1) is a member of the evolutionarily conserved family of ELMO proteins.

Application

Anti-ELMO antibody produced in rabbit is suitable for immunoprecipitation at a working antibody amount of 1-2μL using a whole extract of mouse A-20 cells. It is also suitable for immunoblotting at a working dilution of 1:2000-1:3000 using a whole extract of human Jurkat cells and at 1:1000-1:2000 using a whole extract of rat brain.

Biochem/physiol Actions

ELMO proteins regulate actin cytoskeleton reorganization during engulfment and cell migration. ELMO1 and Dock180 function together as a guanine exchange factor (GEF) for the small GTPase Rac, leading to its activation and cytoskeleton rearrangement. The C-terminal PH domain of ELMO1 binds to Dock180 and the N-terminal binds to the active form of the small GTPase RhoG, targeting the ELMO/Dock180 complex to the membrane. It also interacts with BAI1, an engulfment receptor for apoptotic cells upstream of the ELMO1/Dock180/Rac module. Increased expression of ELMO1 and Dock180 are associated with the invasive phenotype of glioma cells.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

IXO14786: + E1908-200UL: + E1908-25UL: + E1908-BULK: + E1908-VAR:

jan



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters