コンテンツへスキップ
Merck
Dissolution testing of amorphous solid dispersions.
International journal of pharmaceutics (2013-02-02)
K Kogermann, A Penkina, K Predbannikova, K Jeeger, P Veski, J Rantanen, K Naelapää
要旨
The main purpose of this study was to investigate the effect of different polymers, with varying physicochemical properties and molecular weight on the stability and dissolution of co-milled amorphous solid dispersions (ASDs) of piroxicam (PRX). The stability of amorphous PRX (aPRX) in ASDs was significantly improved by the polymers. In-line Raman spectroscopy revealed that solvent mediated solid state changes occurred in biorelevant medium, however differences between ASDs were found. Thus, the dissolution behavior of ASDs of PRX and the respective polymer during conventional large volume (900ml) and a commercial small volume (20ml) dissolution testing was evaluated. The results of these studies indicated that the molecular weight of the polymer (PVP90 vs PVP25) is influencing the solubility of PRX from ASD. Interestingly the effect of molecular weight of the polymer was different than reported previously in the literature for the similar ASDs prepared by spray drying. Furthermore, the dose related bioavailability was determined by investigating the experimental saturation concentrations for different doses. These studies confirmed the findings of the dissolution studies. The differences are presumably caused by the formation of physically different diffusion layers around the ASD particles.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ポリビニルピロリドン, average mol wt 40,000
Sigma-Aldrich
ポリビニルピロリドン, mol wt (number average molecular weight Mn 360kDa)
Sigma-Aldrich
メチルセルロース, viscosity: 4,000 cP
Sigma-Aldrich
ポリビニルピロリドン, powder, average Mw ~55,000
Sigma-Aldrich
ポリビニルピロリドン, average mol wt 10,000
Sigma-Aldrich
ポリビニルピロリドン, average Mw ~1,300,000 by LS
Sigma-Aldrich
ポリビニルピロリドン, K 30
Sigma-Aldrich
ポリビニルピロリドン, K 90
Sigma-Aldrich
メチルセルロース, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
メチルセルロース, viscosity: 1,500 cP
Sigma-Aldrich
ポリビニルピロリドン, powder, average Mw ~29,000
Sigma-Aldrich
ポリビニルピロリドン, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
メチルセルロース, viscosity: 15 cP
Sigma-Aldrich
メチルセルロース, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Kollidon® 25
Sigma-Aldrich
メチルセルロース, viscosity: 25 cP
Sigma-Aldrich
ポリビニルピロリドン 溶液, K 60, 45% in H2O
Sigma-Aldrich
ポリビニルピロリドン, Molecular Biology, nucleic acid hybridization tested, mol wt 360,000
Sigma-Aldrich
メチルセルロース, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Sigma-Aldrich
メチルセルロース, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Supelco
ポリビニルピロリドン, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methocel® A15 LV, 27.5-31.5% methoxyl basis
Sigma-Aldrich
ピロキシカム, ≥98% (TLC)
Sigma-Aldrich
ポリビニルピロリドン, K 25, tested according to Ph. Eur.
Sigma-Aldrich
ポリビニルピロリドン, CP, Mw 1200000, K 90, 11.5-12.8% N basis
Sigma-Aldrich
ポリビニルピロリドン, suitable for plant cell culture, average mol wt 10,000
Sigma-Aldrich
Povidone, meets USP testing specifications
ポリビニルピロリドン, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Methocel® A15C, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
メチルセルロース, tested according to Ph. Eur.