コンテンツへスキップ
Merck
  • The study of fingerprint characteristics of Dayi Pu-Erh tea using a fully automatic HS-SPME/GC-MS and combined chemometrics method.
The study of fingerprint characteristics of Dayi Pu-Erh tea using a fully automatic HS-SPME/GC-MS and combined chemometrics method.
PloS one (2015-01-01)
Shidong Lv, Yuanshuang Wu, Jiangsheng Zhou, Ming Lian, Changwen Li, Yongquan Xu, Shunhang Liu, Chao Wang, Qingxiong Meng
要旨
The quality of tea is presently evaluated by the sensory assessment of professional tea tasters, however, this approach is both inconsistent and inaccurate. A more standardized and efficient method is urgently needed to objectively evaluate tea quality. In this study, the chemical fingerprint of 7 different Dayi Pu-erh tea brands and 3 different Ya'an tea brands on the market were analyzed using fully automatic headspace solid-phase microextraction (HS-SPME) combined with gas chromatography-mass spectrometry (GC-MS). A total of 78 volatiles were separated, among 75 volatiles were identified by GC-MS in seven Dayi Pu-erh teas, and the major chemical components included methoxyphenolic compounds, hydrocarbons, and alcohol compounds, such as 1,2,3-trimethoxybenzene, 1,2,4-trimethoxybenzene, 2,6,10,14-tetramethyl-pentadecane, linalool and its oxides, α-terpineol, and phytol. The overlapping ratio of peaks (ORP) of the chromatogram in the seven Dayi Pu-erh tea samples was greater than 89.55%, whereas the ORP of Ya'an tea samples was less than 79.10%. The similarity and differences of the Dayi Pu-erh tea samples were also characterized using correlation coefficient similarity and principal component analysis (PCA). The results showed that the correlation coefficient of similarity of the seven Dayi Pu-erh tea samples was greater than 0.820 and was gathered in a specific area, which showed that samples from different brands were basically the same, despite have some slightly differences of chemical indexes was found. These results showed that the GC-MS fingerprint combined with the PCA approach can be used as an effective tool for the quality assessment and control of Pu-erh tea.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
ジビニルベンゼン, technical grade, 55%
Sigma-Aldrich
ジビニルベンゼン, technical grade, 80%
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, red hub
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, red hub
Sigma-Aldrich
オクタメチルトリシロキサン, viscosity 1.0 cSt (25 °C)
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, red hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, blue hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, pink hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, blue hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 3, pink hub
Supelco
SPMEファイバーアセンブリー ポリアクリラート (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, white hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, StableFlex (1 cm), needle size 24 ga, Autosampler, pk of 3, pink hub
Supelco
SPMEポータブルフィールドサンプラー, coating PDMS/DVB
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, blue hub
Supelco
SPMEファイバーアセンブリー ポリアクリラート (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, white hub
Supelco
SPMEポータブルフィールドサンプラー, coating PDMS
Supelco
SPME StableFlexファイバー組み合わせキット, Assortment, StableFlex (1 cm), needle size 24 ga, Manual Holder, pk of 4
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 100 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Manual Holder, pk of 3, red hub
Sigma-Aldrich
ポリ(ジメチルシロキサン), viscosity 0.65 cSt (25 °C)
Supelco
SPME StableFlexファイバー組み合わせキット, Assortment, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPMEポータブルフィールドサンプラー, coating CAR/PDMS
Supelco
SPMEファイバー組み合わせキット4, Assortment #4, Fused Silica (1 cm), size 24 ga, Manual Holder, pk of 3
Supelco
SPMEファイバーアセンブリ、Carbowax・ポリエチレングリコール(PEG)コーティング, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Autosampler, pk of 3, purple hub
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 7 μm PDMS, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, green hub
Supelco
SPMEファイバーアセンブリー ポリアクリラート (PA), 85 μm Polyacrylate, Fused Silica (1 cm), needle size 24 ga, Manual Holder, pk of 3, white hub
Supelco
SPMEファイバーアセンブリー ポリジメチルシロキサン(PDMS), 30 μm PDMS, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3, yellow hub
Supelco
SPMEファイバー・ポリジメチルシロキサン/ジビニルベンゼン(PDMS/DVB), 65 μm PDMS/DVB, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 3, blue hub
Supelco
SPME Fiber Assortment Kit 5(SPME用ファイバーアソートメントキット5), Assortment #5, Fused Silica (1 cm), needle size 23 ga, Autosampler, pk of 4
Supelco
SPMEファイバーアセンブリ、Carbowax・ポリエチレングリコール(PEG)コーティング, 60 μm PEG, Metal Alloy (1 cm), needle size 23 ga, Manual Holder, pk of 3, purple hub
Supelco
SPMEファイバー組み合わせキット1, Assortment #1, Fused Silica (1 cm), needle size 24 ga, Autosampler, pk of 3