コンテンツへスキップ
Merck
  • Potential of Piperazinylalkylester Prodrugs of 6-Methoxy-2-Naphthylacetic Acid (6-MNA) for Percutaneous Drug Delivery.
Potential of Piperazinylalkylester Prodrugs of 6-Methoxy-2-Naphthylacetic Acid (6-MNA) for Percutaneous Drug Delivery.
AAPS PharmSciTech (2014-11-06)
Vijay Pawar, Rushabh Thosani, Ashish Kanhed, Rajani Giridhar, Mange Ram Yadav
要旨
Piperazinylalkyl ester prodrugs (4a-5d) of 6-methoxy-2-naphthylacetic acid (6-MNA) (1) were synthesized and evaluated in vitro for the purpose of percutaneous drug delivery. These ionizable prodrugs exhibited varying aqueous solubilities and lipophilicities depending on the pH of the medium. The prodrugs (4a-5c) showed higher aqueous solubility and similar lipophilicity at pH 5.0 and lower aqueous solubility and higher lipophilicity at pH 7.4 in comparison to 6-MNA. The chemical and enzymatic hydrolyses of the prodrugs was investigated in aqueous buffer solutions (pH 5.0 and 7.4) and in 80% human serum (pH 7.4) at 37°C. The prodrugs showed moderate chemical stability (t 1/2 = 6-60 h) but got readily hydrolyzed enzymatically to 6-MNA with half-life ranging from 10-60 min. In the in vitro permeation study using rat skin, the flux of 6-MNA and the prodrugs was determined in aqueous buffers of pH 5.0 and 7.4. The prodrug (5b) showed 7.9- and 11.2-fold enhancement in skin permeation compared to 6-MNA (1) at pH 5.0 and 7.4, respectively. It was concluded that the parent NSAIDs having favorable pharmacokinetic and pharmacodynamic properties coupled with increased skin permeability of their prodrugs could give better options for the treatment of rheumatic diseases.
材料
製品番号
ブランド
製品内容
Sigma-Aldrich
クロロホルム-d, 99.8 atom % D
Sigma-Aldrich
クロロホルム-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
クロロホルム-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
クロロホルム-d, 99.8 atom % D, contains 1 % (v/v) TMS
Sigma-Aldrich
クロロホルム-d, 99.8 atom % D, contains 0.1 % (v/v) TMS
Sigma-Aldrich
クロロホルム-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer
Sigma-Aldrich
クロロホルム-d, ≥99.8 atom % D, contains 0.5 wt. % silver foil as stabilizer, 0.03 % (v/v) TMS
Sigma-Aldrich
臭化カリウム, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
クロロホルム-d, "100%", 99.96 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
クロロホルム-d, "100%", 99.95 atom % D
Sigma-Aldrich
クロロホルム-d, ≥99.8 atom % D, anhydrous
Sigma-Aldrich
臭化カリウム, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
臭化カリウム, anhydrous, powder, 99.95% trace metals basis
Sigma-Aldrich
臭化カリウム, BioXtra, ≥99.0%
Sigma-Aldrich
クロロホルム-d, "100%", 99.96 atom % D, contains 0.5 wt. % silver wire as stabilizer
Sigma-Aldrich
臭化カリウム, JIS special grade, 99.0-100.2%
Sigma-Aldrich
臭化カリウム, SAJ first grade, 99.0-101.0%