コンテンツへスキップ
Merck
Neuroprotective activity of metabotropic glutamate receptor ligands.
Advances in experimental medicine and biology (2003-02-11)
Peter J Flor, Giuseppe Battaglia, Ferdinando Nicoletti, Fabrizio Gasparini, Valeria Bruno
要旨
Metabotropic glutamate receptors form a family of currently eight subtypes (mGluR1-8), subdivided into three groups (I-III). Activation of group-II (mGluR2 and -3) or group-III metabotropic glutamate receptors (mGluR4, -6, -7 and -8) has been established to be neuroprotective in vitro and in vivo. In contrast, group-I mGluRs (mGluR1 and -5) need to be antagonized in order to evoke protection. Initially, all neuroprotective mGluR ligands were analogues of L-glutamate. Those compounds were valuable to demonstrate protection in vitro, but showed limited applicability in animal models, particularly in chronic tests, due to low blood-brain-barrier penetration. Recently, systemically active and more potent and selective ligands became available, e.g., the group-II mGluR agonists LY354740 and LY379268 or group-I antagonists like MPEP (mGluR5-selective) and BAY36-7620 (mGluR1-selective). This new generation of pharmacological agents allows a more stringent assessment of the role of individual mGluR-subtypes or groups of receptors in various nervous system disorders, including ischaemia-induced brain damage, traumatic brain injury, Huntington's and Parkinson's-like pathology or epilepsy. Moreover, the use of genetically modified animals (e.g., knock-out mice) is starting to shed light on specific functions of mGluR-subtypes in experimental neuropathologies.