콘텐츠로 건너뛰기
Merck
  • SKAR is a specific target of S6 kinase 1 in cell growth control.
SKAR is a specific target of S6 kinase 1 in cell growth control.
Current biology : CB (2004-09-03)
Celeste J Richardson, Mark Bröenstrup, Diane C Fingar, Kristina Jülich, Bryan A Ballif, Steven Gygi, John Blenis
초록
The mammalian target of rapamycin (mTOR) and phosphatidylinositol 3-kinase (PI3K) signaling pathways promote cell growth and cell cycle progression in response to nutritional, energy, and mitogenic cues. In mammalian cells, the ribosomal protein S6 kinases, S6K1 and S6K2, lie downstream of mTOR and PI3K, suggesting that translational control through the phosphorylation of S6 regulates cell growth. Interestingly, genetic experiments predict that a substrate that is specific to S6K1 but not S6K2 regulates cell growth. Here we identify SKAR as a novel and specific binding partner and substrate of S6K1 but not S6K2. We find that serines 383 and 385 of human SKAR are insulin-stimulated and rapamycin-sensitive S6K1 phosphorylation sites. Quantitative mass spectrometry reveals that serine 383/385 phosphorylation is sensitive to RNA interference (RNAi)-mediated S6K1 reduction, but not S6K2 reduction. Furthermore, RNAi-mediated reduction of SKAR decreases cell size. SKAR is nuclear protein with homology to the Aly/REF family of RNA binding proteins, which has been proposed to couple transcription with pre-mRNA splicing and mRNA export. We have identified a novel and specific target of S6K1, SKAR, which regulates cell growth. The homology of SKAR to the Aly/REF family links S6K1 with mRNA biogenesis in the control of cell growth.