Merck
Structure and genetics of the O-antigen of Enterobacter cloacae G3054 containing di-N-acetylpseudaminic acid.
Carbohydrate research (2015-02-28)
Andrei V Perepelov, Min Wang, Andrei V Filatov, Xi Guo, Alexander S Shashkov, Lei Wang, Yuriy A Knirel
RESUMEN
Mild acid degradation of the lipopolysaccharide of Enterobacter cloacae G3054 resulted in the cleavage of the O-polysaccharide at the linkage of residues of 5,7-diacetamido-3,5,7,9-tetradeoxy-L-glycero-L-manno-non-2-ulosonic acid (di-N-acetylpseudaminic acid, Pse5Ac7Ac) in the main chain. The resultant oligosaccharide and an alkali-treated lipopolysaccharide were studied by sugar analysis along with (1)H and (13)C NMR spectroscopy, and the following structure of the branched pentasaccharide O-unit of the O-polysaccharide was established: [structure: see text] The O-antigen gene cluster of E. cloacae G3054 between conserved genes galF and gnd was sequenced. Most genes necessary for the O-antigen synthesis were found in the cluster and their functions were tentatively assigned by comparison with sequences in the available databases.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glicerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glicerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glicerol, ≥99.5%
Sigma-Aldrich
Glicerol solution, 83.5-89.5% (T)
Sigma-Aldrich
Glicerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glicerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
Glicerol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glicerol, BioXtra, ≥99% (GC)
USP
Glicerol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Glicerol, puriss., anhydrous, 99.0-101.0% (alkalimetric)
Sigma-Aldrich
Glicerol, FCC, FG
Sigma-Aldrich
Glicerol, puriss. p.a., ACS reagent, anhydrous, dist., ≥99.5% (GC)
Sigma-Aldrich
Glicerol, meets USP testing specifications
Sigma-Aldrich
Glicerol solution, puriss., meets analytical specification of Ph. Eur., BP, 84-88%
Supelco
Glicerol, analytical standard
Sigma-Aldrich
Glicerol, tested according to Ph. Eur., anhydrous