Merck
  • Role of imidazoline receptors in the cardiovascular actions of moxonidine, rilmenidine and clonidine in conscious rabbits.
Role of imidazoline receptors in the cardiovascular actions of moxonidine, rilmenidine and clonidine in conscious rabbits.
The Journal of pharmacology and experimental therapeutics (1996-02-01)
C K Chan, F Sannajust, G A Head
ABSTRACT
The present study in conscious rabbits with intracisternal (i.c.) catheters sought to determine the relative contribution of the I1 subtype of imidazoline receptors (IR) and alpha 2 adrenoceptors to the hypotensive effects of rilmenidine, clonidine and moxonidine with an I1-IR/alpha 2 adrenoceptor antagonist efaroxan and a specific alpha 2 adrenoceptor antagonist 2-methoxyidazoxan (2-MI). The alpha 2 adrenoceptor antagonist effect of efaroxan was compared with 2-MI by performing cumulative dose-response curves in the presence of alpha-methyldopa (400 micrograms/kg i.c.). 2-MI was 5.6 times more potent than efaroxan at reversing 75% of the hypotension elicited by alpha-methyldopa (P < .025). This dose ratio was used to match doses of efaroxan and 2-MI for similar alpha 2 adrenoceptor blockade. The effects of efaroxan (4.1, 13, 41 micrograms/kg i.c.) and 2-MI (0.74, 2.3, 7.4 micrograms/kg i.c.) were investigated on a single i.c. dose of rilmenidine (12 micrograms/kg), clonidine (0.75 microgram/kg) and moxonidine (0.51 microgram/kg). These doses of the antihypertensive agents, which were determined from cumulative dose-response curves, produce 90% of the maximum hypotension. Efaroxan was more effective at reversing the hypotension induced by moxonidine and rilmenidine than was 2-MI (P < .01). These findings suggest that rilmenidine and moxonidine act predominantly via IR. By contrast, 2-MI was more effective at reversing the clonidine-induced hypotension than was efaroxan (P < .001), suggesting that clonidine acts mainly via alpha 2 adrenoceptors in conscious normotensive rabbits. Thus, a higher selectivity of the second generation agents moxonidine and rilmenidine for I1-IR over alpha 2 adrenoceptors, compared with the first generation agent clonidine, appears to be necessary for this effect to be manifested in their hypotensive actions.
Sigma-Aldrich
Rilmenidine hemifumarate salt, solid
Pricing and availability is not currently available.