Merck
Large surface relaxation in the organic semiconductor tetracene.
Nature communications (2014-11-08)
Hazuki Morisaki, Takashi Koretsune, Chisa Hotta, Jun Takeya, Tsuyoshi Kimura, Yusuke Wakabayashi
ABSTRACT
Organic crystals are likely to have a large degree of structural relaxation near their surfaces because of the weak inter-molecular interactions. The design of organic field-effect transistors requires a detailed knowledge of the surface relaxation as the carriers usually transfer within the first few molecular layers at the semiconductor surfaces, and their transport properties reflect the structural changes through the transfer integral. Here, we report the direct observation of the surface relaxation of an organic semiconductor, a tetracene single crystal, by means of X-ray crystal truncation rod scattering measurements. A significant degree of surface relaxation is observed, taking place only in the first monolayer at the semiconductor surface. First principles calculations show that the resultant transfer integrals are completely different between the bulk and surface of the semiconductor.
Sigma-Aldrich
Benz[b]anthracene, sublimed grade, 99.99% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Benz[b]anthracene, 98%
Pricing and availability is not currently available.