Sign In to View Organizational & Contract Pricing
Select a Size
Pricing and availability is not currently available.
Skip To
material
acetal (polypropylene) housing
cellulose filter aid (and Diatomaceous earth (DE))
Quality Level
product line
Millistak+®
feature
hydrophilic
filtration area
1.1 m2
impurities
(LAL test)
input
liquid
fitting
fitting Flat Seal
application(s)
pharma/biopharma processes
code
D0HC
shipped in
ambient
General description
To improve sustainability of the packaging and shipping of filter products, Optimized Pack solutions for Millistak+® HC Process Scale Pod filters have been developed to optimize transport and receiving processes as well as to reduce waste.
Application
Primary clarification.
Features and Benefits
The key benefits are:
- 24% reduction in corrugated packaging waste to recycle or dispose.
- 94% reduction of paper documentation.
- 12% decrease in number of deliveries which will further reduce energy use and emissions.
- 70% reduction in operator time to open and manage the product and packaging.
Packaging
36/pack Optimized packaging configuration includes 36 pods distributed in 3 boxes on one APAC standardized pallet (pallet size: 1100*1100mm).
Other Notes
Integrity is maintained after 1 autoclave cycle of 60 minutes at 123 °C
Legal Information
MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany
related product
Product No.
Description
Pricing
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service