Skip to Content
Merck

EMU026171

MISSION® esiRNA

targeting mouse Sh3pxd2a

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAGAAGGATCCGAAGCAAAGGATTATTCCCTTTCTTCCAGGCAAGATCCTCTTCCGGAGAAGCCACATCCGCGACGTGGCTGTGAAGAGACTAAAGCCCATCGATGAATACTGCAGGGCGCTTGTCCGGCTGCCGCCCCACATTTCACAGTGTGACGAAGTCTTCCGGTTCTTTGAGGCACGGCCTGAGGATGTCAACCCTCCAAAAGAAGACTATGGCAGTTCCAAGAGGAAATCAGTGTGGTTGTCCAGCTGGGCTGAGTCTCCCAAGAAGGACGTGACAGGTGCCGACACCAACGCCGAGCCCATGATCCTGGAACAGTACGTGGTGGTGTCCAACTATAAGAAACAAGAGAACTCGGAGCTGAGCCTCCAGGCCGGGGAGGTGGTAGATGTCATCGAGAAGAACGAAAGCGGCTGGTGGTTTGTGAGCACATCTGAAGAGCAAGGTTGGGTC

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
EMU070071EMU021811EMU018671
product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

Ensembl | mouse accession no.

ENSMUSG00000053617

Ensembl | mouse accession no.

ENSMUSG00000040711

Ensembl | mouse accession no.

ENSMUSG00000026193

Ensembl | mouse accession no.

ENSMUSG00000030357

esiRNA cDNA target sequence

CAGAAGGATCCGAAGCAAAGGATTATTCCCTTTCTTCCAGGCAAGATCCTCTTCCGGAGAAGCCACATCCGCGACGTGGCTGTGAAGAGACTAAAGCCCATCGATGAATACTGCAGGGCGCTTGTCCGGCTGCCGCCCCACATTTCACAGTGTGACGAAGTCTTCCGGTTCTTTGAGGCACGGCCTGAGGATGTCAACCCTCCAAAAGAAGACTATGGCAGTTCCAAGAGGAAATCAGTGTGGTTGTCCAGCTGGGCTGAGTCTCCCAAGAAGGACGTGACAGGTGCCGACACCAACGCCGAGCCCATGATCCTGGAACAGTACGTGGTGGTGTCCAACTATAAGAAACAAGAGAACTCGGAGCTGAGCCTCCAGGCCGGGGAGGTGGTAGATGTCATCGAGAAGAACGAAAGCGGCTGGTGGTTTGTGAGCACATCTGAAGAGCAAGGTTGGGTC

esiRNA cDNA target sequence

CTCCAGATGCAGATGTTGGACAAGTTCCCCATGGAAGGAGGACAGAAAGACCCCAAGCAGAGGATCATTCCATTTCTGCCAGGCAAAATCCTCTTCCGCCGCAGCCACATCCGGGACGTAGCCGTCAAACGCCTGATACCAATTGATGAATATTGTAAGGCCCTGATCCAGCTGCCCCCCTACATCTCTCAGTGTGATGAGGTTCTGCAGTTTTTCGAGACAAGACCTGAGGACCTGAATCCTCCCAAAGAGGAGCATATTGGGAAGAAGAAATCAGGGAATGACCCGACCTCAGTGGACCCTATGGTCCTGGAGCAGTATGTGGTAGTGGCAGACTATCAGAAGCAGGAGAGCTCAGAGATCAGCCTCAGTGTGGGCCAAGTAGTGGACATCATTGAGAAGAATGAGTCAGGTTGGTGGTTCGTC

esiRNA cDNA target sequence

AGTGCTTCATGCCGCTAGATGTGCAAGCTGACAGAGACGATTCTCGAGAGTAATCTTTCCAGCCCCACCCTACAAGTGTCTCTCTACCAAGGTCAATCCACACCCCAGTGATGTTAGCAGACCCTCCATCTTTGAGTGGTCCTTTCACCCTTAAGCCTTTTGCTCTGGAGCCATGTTCTCAGCTTCAGCACAATTTACAGCTTCTCCAAGCATCGCCCCGTGGGATGTTTTGAGACTTCTCTCCTCAATGGTGACAGTTGGTCACCCTGTTCTGCTTCAGGGTTTCAGTACTGCTCAGTGTTGTTTAAGAGAATCAAAAGTTCTTATGGTTTGGTCTGGGATCAATAGGGAAACACAGGTAGCCAACTAGGAGGAAATGTACTGAATGCTAGTACCCAAGACCTTGAGCAGGAAAGTCACCCAGACACCTCTGCTTTCTTTTGCC

esiRNA cDNA target sequence

AGTGTGGGGAAGGAGAGGTTCCAGATCCCACCGCACGCTGAGCTGAGGTATGAAGTGCGGCTGAAGAGCTTTGAGAAGGCCAAGGAGTCTTGGGAGATGAGCTCCGCGGAGAAGCTGGAGCAGAGCAACATAGTGAAAGAGAGGGGCACCGCGTACTTCAAGGAAGGCAAGTACAAGCAGGCGTTACTGCAGTACAAGAAGATCGTGTCTTGGCTAGAATACGAGTCTAGCTTCTCCGGTGAGGAAATGCAAAAGGTCCATGCACTCCGACTGGCCTCACACCTCAATCTGGCCATGTGTCACCTGAAACTGCAGGCCTTCTCAGCTGCCATCGAAAGCTGCAACAAGGCCTTGGAGCTGGACAGCAACAACGAGAAGGGCCTGTTTCGCCGGGGAGAGGCCCACCTGGCCGTGAATGACTTTGACCTGGCAAGAGC

Gene Information

mouse ... SH3PXD2A(14218), Sh3pxd2a(14218)

Gene Information

mouse ... SH3PXD2B(268396), Sh3pxd2b(268396)

Gene Information

mouse ... FN1(14268), Fn1(14268)

Gene Information

mouse ... FKBP4(14228), Fkbp4(14228)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_008018

NCBI accession no.

NM_177364

NCBI accession no.

NM_010233

NCBI accession no.

NM_010219

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Roh-Johnson et al.
Oncogene, 33(33), 4203-4212 (2013-09-24)
Most cancer patients die as a result of metastasis, thus it is important to understand the molecular mechanisms of dissemination, including intra- and extravasation. Although the mechanisms of extravasation have been vastly studied in vitro and in vivo, the process

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service