Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Product Name
54SCC7, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.
biological source
Mouse x Rat Hybridoma nerve
manufacturer/tradename
ECACC
packaging
tube of 5 μg 08062518-DNA-5UG, pkg of vial of cells 08062518-1VL
growth mode
Adherent
karyotype
Modal No. varied with passage see reference
morphology
Oligopolar
products
Not specified
receptors
Not specified
technique(s)
cell culture | mammalian: suitable
relevant disease(s)
cancer
shipped in
dry ice
storage temp.
−196°C
Biochem/physiol Actions
Preparation Note
Other Notes
1 of 1
This Item | |||
|---|---|---|---|
| description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. | description NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA. |
| storage temp. −196°C | storage temp. −196°C | storage temp. −196°C | storage temp. −196°C |
| receptors Not specified | receptors Not specified | receptors Not specified | receptors - |
| karyotype Modal No. varied with passage see reference | karyotype Modal No. varied with passage see reference | karyotype - | karyotype modal No. of chromosomes is 42 |
| packaging pkg of vial of cells 08062518-1VL, tube of 5 μg 08062518-DNA-5UG | packaging tube of 5 μg 08062561-DNA-5UG, pkg of vial of cells 08062561-1VL | packaging - | packaging tube of 5 μg 08062563-DNA-5UG, pkg of vial of cells 08062563-1VL |
| growth mode Adherent | growth mode Adherent | growth mode Adherent | growth mode - |
| morphology Oligopolar | morphology oligopolar | morphology - | morphology - |
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
