Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 979,00 PLN |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 1.760,00 PLN |
About This Item
979,00 PLN
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CAAAGCCAGTTCTCACACCTTTTTCAGATCAAATGTTCCAGCCTCTTCAGAAAAGCTTAGAAAATAGTCTTTCTTTCACTCCCCACCAGTCTTCATTGTGACATATGTTGCATATCATAACCCTTGGTACTGGATGAAAGCTCCCAATCTGAATCAAGAACAGGTAAAGGTAAACAAAAAATTGAGGCTAAAAGAATCACTTAACCTGAACCACATGGTAGGAAAAGCATGCTAGAATGGAATATCCAACCGGCTATGGACATTGTTTTTGGCAGACAAATGAATGGCCCCATTATAAAGGCATCAGGGTTGGTGTATTGTCCAACTCTAGGGGGCAACATTGAATTGTATCCTATCGTTATGGTGCTCCCTGGAGTTCTGTAAACACATAGGTCTGAGAGTGATTATCACATGCCTATCTTATCCAGGGAAATAAGGAGCAAAATGGCCCAGAAGAAGAGAAAGAACATTCTACCAAGTCACACGTAACCTACCAACAGTCCACAACTGCAATTGATTTCATGCCCAACTCTTGC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... APIP(51074)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting human APIP | description targeting mouse Apip | description targeting human APIP (2) | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, ab2 |
| esiRNA cDNA target sequence CAAAGCCAGTTCTCACACCTTTTTCAGATCAAATGTTCCAGCCTCTTCAGAAAAGCTTAGAAAATAGTCTTTCTTTCACTCCCCACCAGTCTTCATTGTGACATATGTTGCATATCATAACCCTTGGTACTGGATGAAAGCTCCCAATCTGAATCAAGAACAGGTAAAGGTAAACAAAAAATTGAGGCTAAAAGAATCACTTAACCTGAACCACATGGTAGGAAAAGCATGCTAGAATGGAATATCCAACCGGCTATGGACATTGTTTTTGGCAGACAAATGAATGGCCCCATTATAAAGGCATCAGGGTTGGTGTATTGTCCAACTCTAGGGGGCAACATTGAATTGTATCCTATCGTTATGGTGCTCCCTGGAGTTCTGTAAACACATAGGTCTGAGAGTGATTATCACATGCCTATCTTATCCAGGGAAATAAGGAGCAAAATGGCCCAGAAGAAGAGAAAGAACATTCTACCAAGTCACACGTAACCTACCAACAGTCCACAACTGCAATTGATTTCATGCCCAACTCTTGC | esiRNA cDNA target sequence GCCAGTGCACTCCTCTTTTCATGAATGCTTATACCATGAGAGGAGCTGGCGCAGTGATTCATACCCACTCTAAAGCTGCTGTGATGGCTACCCTTCTGTTTCCAGGACAGGAGTTTAAAATTACACATCAAGAGATGATCAAAGGAATAAGGAAATGTACCTCAGGAGGCTATTACAGATACGATGATATGTTAGTGGTACCTATTATTGAGAACACTCCTGAAGAGAAGGATCTCAAAGAAAGGATGGCTCATGCCATGAACGAGTACCCAGACTCCTGTGCGGTTCTTGTCCGGCGTCATGGGGTGTACGTGTGGGGAGAAACATGGGAGAAAGCAAAAACCATGTGTGAGTGTTATGACTACCTGTTTGACATTGCTGTCTCCATGAAGAAGATGGGACTCGATCCAA | esiRNA cDNA target sequence ATGAGAGGAGCAGGTGCAGTGATTCATACCCACTCTAAAGCTGCTGTGATGGCCACACTTCTCTTTCCAGGACGGGAGTTTAAAATTACACATCAAGAGATGATAAAAGGAATAAAGAAATGTACTTCCGGAGGGTATTATAGATATGATGATATGTTAGTGGTACCCATTATTGAGAATACACCTGAGGAGAAAGACCTCAAAGATAGAATGGCTCATGCAATGAATGAATACCCAGACTCCTGTGCAGTACTGGTCAGACGTCATGGAGTATATGTGTGGGGGGAAACATGGGAGAAGGCCAAAACCATGTGTGAGTGTTATGACTATTTATTTGATATTGCCGTATCAATGAAGAAAGTAGGACTTGATCCTTCACAGCTCCCAGTTGGAGAAAATGGAATTGTCTAAGCCAAAAGAAAGTCTAATTATATACAGAGATAAAGCTAAACGTAATTATTATTTAAATGAAAGCTATTTTTTTAAATGAATTGAAATTTTTCATGATGCTACTAATTTGCCACTAAATACTGCAAATGGTCACCCTGAATC | esiRNA cDNA target sequence - |
| product line MISSION® | product line MISSION® | product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies |
| Gene Information human ... APIP(51074) | Gene Information mouse ... APIP(56369), Apip(56369) | Gene Information human ... APIP(51074) | Gene Information human ... APIP(51074) |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form buffered aqueous glycerol solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| GenBank accession no. | GenBank accession no. - | GenBank accession no. - | GenBank accession no. - |
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
WGK
WGK 1
Storage Class
12 - Non Combustible Liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

