Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 979,00 PLN |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 1.760,00 PLN |
About This Item
979,00 PLN
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TCCGAAGAGAGGTGGAGAAACCTAAAGACTGAGGAGAAGAGATCCTTTGAGCCAGATGGGGCATTAGTTCTTCTGCTTTTCTCAGCATGGATAAACCATTTCCTCAAGGATTTTCTCATGTGCCCTGAAATCCATGTAACTACAAGGGCTCCTCTTTATCACCATAAGTGCCACCCTGACTTAAAACCACTCAGAGCTAAAAAATCAAGGCAAAATGGATGCTGCGGTGACAGATGATTTTCAACAAATTCTGCCTATTGAACAGCTGCGCTCTACTCATGCTAGCAATGACTACGTGGAACGGCCTCCAGCCCCCTGTAAACAGGCCCTCTCCAGCCCTTCCCTTATTGTGCAAACCCACAAGTCTGATTGGTCTCTGGCTACCATGCCTACTTCTCTCCCCCGCAGTCTCAGCCAGTGCCATCAACTGCAGCCCTTGCCTCAGCATCTGAGCCAATCTAGCATTGCCAGCTCAATGTCCCATAGCACCACTGCCTCTGA
Ensembl | human accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
human ... SPRY3(10251)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | ||
|---|---|---|
| description targeting human SPRY3 | description affinity isolated antibody | description targeting mouse Spry3 |
| esiRNA cDNA target sequence TCCGAAGAGAGGTGGAGAAACCTAAAGACTGAGGAGAAGAGATCCTTTGAGCCAGATGGGGCATTAGTTCTTCTGCTTTTCTCAGCATGGATAAACCATTTCCTCAAGGATTTTCTCATGTGCCCTGAAATCCATGTAACTACAAGGGCTCCTCTTTATCACCATAAGTGCCACCCTGACTTAAAACCACTCAGAGCTAAAAAATCAAGGCAAAATGGATGCTGCGGTGACAGATGATTTTCAACAAATTCTGCCTATTGAACAGCTGCGCTCTACTCATGCTAGCAATGACTACGTGGAACGGCCTCCAGCCCCCTGTAAACAGGCCCTCTCCAGCCCTTCCCTTATTGTGCAAACCCACAAGTCTGATTGGTCTCTGGCTACCATGCCTACTTCTCTCCCCCGCAGTCTCAGCCAGTGCCATCAACTGCAGCCCTTGCCTCAGCATCTGAGCCAATCTAGCATTGCCAGCTCAATGTCCCATAGCACCACTGCCTCTGA | esiRNA cDNA target sequence - | esiRNA cDNA target sequence TGGTAGCTGCCTAAGAACTTAGATTTCAGCAACAGTGAACAACTGAGATGAAAGATATGTAAAGCACAGAAGTGACTCCCCTCATAAAATGTACAAAGAGCCTGTGCTACGAACCCCTGTCAATAAGGAAAATGCTGCAATGGTGATGGCAGGGTGCAAAAGACTGCTACTAAGAGACATGAGAATTATATAGTTTCCCTCTGTAAGTAAACCCAAAATTTATCAAGGAATACAGATATTGGGGGCACTTGCTTGAAATCAAGGTGCTCCAACTTAGTTTAAGACTTCCAGACTCTATGTATCACCTCTTTTAAAGTATGCATGGATGTGTGCTGCAGGGTGAAGTAAGGAGAAGTGCATAATCATGCACAAGAGGAAGAAAGGGAATTACATGTCCCTTTATTGGAAATATACTGCAGAAATATATGCCACAAAGTATTTATTGGCTATGAAACTTCCTGAGGTGTATTGACATTGGGTTGCACACAGCCACATACCTTCACTTACATCTCC |
| Gene Information human ... SPRY3(10251) | Gene Information human ... SPRY3(10251) | Gene Information mouse ... SPRY3(236576), Spry3(236576) |
| Ensembl | human accession no. | Ensembl | human accession no. - | Ensembl | human accession no. - |
| product line MISSION® | product line - | product line MISSION® |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form buffered aqueous solution | form lyophilized powder |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

