Skip to Content
Merck

Key Documents

Skip To

EHU080751

MISSION® esiRNA

targeting human MON2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
979,00 PLN
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
1.760,00 PLN

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

979,00 PLN


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

ATGGAAATACCTGGGCACAAGTAATTGCCTTATACCCAACTTTAGTAGAATGCATCACCTGTTCTTCTTCAGAAGTCTGTTCTGCACTTAAAGAGGCACTAGTTCCTTTTAAGGATTTCATGCAGCCACCAGCATCCAGAGTTCAAAATGGAGAATCTTGACCGGCTACAATATATTTGAAAGCAGGAAGATAGTCTAAAAAATGTTTGCTCCTAATTGAGTCTTCTGTGAGAAGGACATTTCTTACTGCAGATAATTCTTGGCAGCTGTTGTTGGCCTCCTTTAAATTCTACTTACCTGAGTTCAGTAATTCATATTACAGGCTTGCACATCAACAAAGGCTCCTGAATGAACAGCAGTGTAAGGCTTTAATAAATTAAACTGATGGGAGGGATAATTAACACTACAGTATACATGCTACCATATCTCCAGTTGGTGA

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... MON2(23041)

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA038697HPA057607EMU002601
description

targeting human MON2

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

targeting mouse Mon2

Ensembl | human accession no.

ENSG00000061987

Ensembl | human accession no.

-

Ensembl | human accession no.

-

Ensembl | human accession no.

-

Gene Information

human ... MON2(23041)

Gene Information

human ... MON2(23041)

Gene Information

human ... MON2(23041)

Gene Information

mouse ... MON2(67074), Mon2(67074)

product line

MISSION®

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

MISSION®

esiRNA cDNA target sequence

ATGGAAATACCTGGGCACAAGTAATTGCCTTATACCCAACTTTAGTAGAATGCATCACCTGTTCTTCTTCAGAAGTCTGTTCTGCACTTAAAGAGGCACTAGTTCCTTTTAAGGATTTCATGCAGCCACCAGCATCCAGAGTTCAAAATGGAGAATCTTGACCGGCTACAATATATTTGAAAGCAGGAAGATAGTCTAAAAAATGTTTGCTCCTAATTGAGTCTTCTGTGAGAAGGACATTTCTTACTGCAGATAATTCTTGGCAGCTGTTGTTGGCCTCCTTTAAATTCTACTTACCTGAGTTCAGTAATTCATATTACAGGCTTGCACATCAACAAAGGCTCCTGAATGAACAGCAGTGTAAGGCTTTAATAAATTAAACTGATGGGAGGGATAATTAACACTACAGTATACATGCTACCATATCTCCAGTTGGTGA

esiRNA cDNA target sequence

-

esiRNA cDNA target sequence

-

esiRNA cDNA target sequence

GCAATGGATATGGCCTATGGAAATAATAAGGAGCCATCTCTCTTTGCTGTTGCCAAACTATTAGAAACTGGTCTTGTTAATATGCACAGGATAGAGATTTTATGGAGACCTCTGACTGGCCATCTACTTGAGGTCTGCCAGCATCCAAACTCTCGGATGCGGGAGTGGGGAGCAGAAGCTTTGACGTCACTCATTAGAGCGGGGCTGACGTTCAGCCATGAGCCTCCACTTCCACAGAACCAGAGACTACAGCTGCTTTTATTGAACCCATTAAAAGAGATGTCCAATATTAACCATCCAGATATTCGACTCAAGCAATTAGAATGTGTGTTGCAGATTCTGCAAAGCCAGGGAGACAGTCTTGGGCCTGGGTGGCCGTTAGTTCTTGGAGTCATGGGAGCAAT

NCBI accession no.

NM_015026

NCBI accession no.

-

NCBI accession no.

-

NCBI accession no.

NM_153395

form

lyophilized powder

form

buffered aqueous glycerol solution

form

buffered aqueous glycerol solution

form

lyophilized powder


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters