Skip to Content
Merck

Skip To

EMNC010161

MISSION® esiRNA

targeting mouse Lpp

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
979,00 PLN
50 μg
Usually ships in 3 weeks (4 to 6 weeks for custom orders).
1.760,00 PLN

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

979,00 PLN


Usually ships in 3 weeks (4 to 6 weeks for custom orders).

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCCTCTGCGAGACTGATTTCAAATCTGTGATCTTCTGTTCAGCGTACACATCAGCAAAGTGAGAAGTTGGGTGCTAAATATAGGCTCTATTAACTTTACTTTTAGATGTGCTGCCTTCAAAGAGTGCCTATTCTGAGCAACGTAAACGTTATTCCTACATATGTATGTACACACGGTACCCGGACTCACACTGTTCGGGCTTCACAAACACCATCAGCAGGGGTAG

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU174181EHU012541SAB5701216
description

targeting mouse Lpp

description

targeting mouse Lpp

description

targeting human LPP

description

-

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

-

esiRNA cDNA target sequence

GCCTCTGCGAGACTGATTTCAAATCTGTGATCTTCTGTTCAGCGTACACATCAGCAAAGTGAGAAGTTGGGTGCTAAATATAGGCTCTATTAACTTTACTTTTAGATGTGCTGCCTTCAAAGAGTGCCTATTCTGAGCAACGTAAACGTTATTCCTACATATGTATGTACACACGGTACCCGGACTCACACTGTTCGGGCTTCACAAACACCATCAGCAGGGGTAG

esiRNA cDNA target sequence

CAAAACAAAAACCATGATTCTCTCTGTCAGCCCAAGAGTGTGCTGAGGCAACCAGGAGTTTCTAATCAAACAAACCCAAGTCCAGAGGCCTTGAAATGTCTAAACAAGTGCTATTGAATGAACAGACCTCCTGAAATGTACCCTTCATTGTATCTATGGGATTAATCTACCCTCATCACAAGGAGCATGACACTGTATTCTTAGCACCAACAAAAACCCTGCTTGGAGCACCCTGGGTACATTCTAGAAGTCAGCCGACATTGCAGGTCAACAAATCAACACTCTTCTTAGAGCTGACTTCCTCGTGAATATGCCTTCCAGGTGCAGTCTTGGCTTTCCTTTTTCTATCTTCTGGTTGTTTGCTACAAGTTGAAAATCCGTTGACTAATTAAGAAAATATTCTCTGTCCACATTAAATCTGTCCTCAATCTAGAAACCCTTTCAAATGCCCTAGTTTGTAAGTTAGGCATTTGTCTTCCCTAGAGCCTCTCCTCCATCAGACCAAGATTACCG

esiRNA cDNA target sequence

TGCATCATCTGCAACAACAAGCTCCGAGGGCAGCCATTCTATGCTGTGGAAAAGAAAGCATACTGCGAGCCCTGCTACATTAATACTCTGGAGCAGTGCAATGTGTGTTCCAAGCCCATCATGGAGCGGATTCTCCGAGCCACCGGGAAGGCCTATCATCCTCACTGTTTCACCTGCGTGATGTGCCACCGCAGCCTGGATGGGATCCCATTCACTGTGGATGCTGGCGGGCTCATTCACTGCATTGAGGACTTCCACAAGAAATTTGCCCCGCGATGTTCTGTGTGCAAGGAGCCTATTATGCCAGCCCCGGGCCAGGAGGAGACTGTCCGTATTGTGGCTTTGGATCGAGATTTCCATGTTCACTGCTACCGATGCGAGGATTGCGGTGGTCTCCTGTCTGAAGGAGATAACCAAGGCTGCTACCCC

esiRNA cDNA target sequence

-

Gene Information

mouse ... LPP(210126), Lpp(210126)

Gene Information

mouse ... LPP(210126)

Gene Information

human ... LPP(4026)

Gene Information

human ... LPP(4026)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_001145952.1, NM_178665.5, NM_001145954.1

NCBI accession no.

-

NCBI accession no.

NM_005578

NCBI accession no.

-

GenBank accession no.

AK034659

GenBank accession no.

-

GenBank accession no.

-

GenBank accession no.

-


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters