Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 979,00 PLN |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 1.760,00 PLN |
About This Item
979,00 PLN
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
ACTGTGTGTGGTCCCTGTTGCTGACAGTATTAAGTTATATTCTGATGTAAGATTAACTTTATTAAAGAATGTAAACATTAATGTTTCCTTATGGGAAAACAAATAAAGTATAAAGAAGACAATTCTTTTTATTGAAATATACTGTGTATTTACACTTGCTAGACCCAGCACCACTTATAAACTTAGTCCACTGTCCAGTTTCAGTTAACACAGCTGACACGGTTGT
GenBank accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... IGIP(109169), 6330403M23Rik(109169)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting mouse Igip | description targeting human AC016735.2 | description targeting human AC069404 | description targeting human RP11-415J8.5 |
| esiRNA cDNA target sequence ACTGTGTGTGGTCCCTGTTGCTGACAGTATTAAGTTATATTCTGATGTAAGATTAACTTTATTAAAGAATGTAAACATTAATGTTTCCTTATGGGAAAACAAATAAAGTATAAAGAAGACAATTCTTTTTATTGAAATATACTGTGTATTTACACTTGCTAGACCCAGCACCACTTATAAACTTAGTCCACTGTCCAGTTTCAGTTAACACAGCTGACACGGTTGT | esiRNA cDNA target sequence TTTGTGAGGATGGCTTTGTGTGGCTGCACATTGCCCTTGGCCAAGCCCCGCTGCCAGGAGAGGCAGCTCGGCTGCCAAACCTCCTGAACCCAAGTTACCTGAGCAAGCAGCCCTATCCCCTCGCCTGCTTCTCACTGCCTCGCCTGCCTTGGGCCTGCCCGACCCAGGGGGTCACATGCCTTGTTCCCCATCAGTGGAGACCAGGGACAAGAAGACAGTTCTGTGGGAAGAGCCAGGAGGACCATTTGGGAAAATACAGGTTCCAGGTGTGTCTCCAGACCTTGAGAGTTGGTGTCTCCTGGGAAGAGGCCCTGATGAGATCTGGGGAGATTCGGATCTGATGAGATTCAGGAAGCCCTGGCCTCTCCGTACTTGAGGAGACGGGACACACAGGACAAGCTGCAG | esiRNA cDNA target sequence GCTGGGTGAATTCTGCTTACAGTGTTTTCCAAAGAGGCCTGTAGGACATCTGGGTCTTCCATGGTGGTGGGGGTTTTGGCCTCAACAACAAGGCTCTGGTGGCAGAAATGATAGCAACCCAGTGAAAAGAGAGGGACAAAAGGGTCAAAGGAGTCAGTGCAAAAGCACTGTCTGGGAAAGCGAATCAACCCAGATTGTCCCCTCGGGAAACATGGCATGTGCTTGGTGTGACAGCTGATTCTCCCCTTAATCACTGCCATGAAAACAAAATAACCATAAGAACAATGGCAGATTAGACAAGGGAAATTAAGGAACCCAAATACTGACAGGACGCATTTTTGAAGATACAGCAGACTCCTGGGTCCTTGTGACAACCTTCCCACACCAATTCTCAGACCACCTTCTACCCGAACCTGCACGGTGTCAAAATGGCCTCCACAAGAAGCAATCCAATGGGGCTCTGTTCTCTCCTTGCCAAGACCACGATGATAACGCAGTTGTGATGGCCACAGGACTAGGAAGGACCATGGA | esiRNA cDNA target sequence GGTTGAGGCTGATTGAGGAAGTGGGTTCAGGCGGCCCGCCGTGCCTGGCACACGGTCAGAGTCCCCTGACAGGACTTTCCCCTTTCTCTGAGATGACAATCAGGCCGCTTTGCTTTGCACTGCCCTCAGAGAAGCAGTGTGCCCAGAAGTCAAGTGTGAGCTCTTGAGTCAAAATGCTTGGATTTGAATTCCAACTCTGCTGCTTACCAGCTGTGTGACATGGGGTGAGTGAGTTACCTGACCTCTCTGTACCTCTATGTCATCATCTTTAAAATGGAGCCCACAGCAACCAGGGAAATAGTCTAACAGAAGCCTAGGATGAAGAATTTCGGGAAAGAAGAAACGTCCAGCAAGGCCAAGTCCCTTTGTGTGTCAGTCTGCTTTCTCTCTCACACATACGTGCACACAGGCTCGTACACAC |
| Gene Information mouse ... IGIP(109169), 6330403M23Rik(109169) | Gene Information - | Gene Information - | Gene Information - |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| NCBI accession no. | NCBI accession no. - | NCBI accession no. - | NCBI accession no. - |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
