Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 979,00 PLN |
50 μg | Usually ships in 3 weeks (4 to 6 weeks for custom orders). | 1.760,00 PLN |
About This Item
979,00 PLN
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
GTGGGATATCGGTGGTCAAGAATCTCTGAGGTCATCCTGGAACACCTATTACTCAAACACAGAGTTCATCATTCTTGTTGTGGATAGCATTGACAGGGAACGCCTGGCTATTACGAAAGAAGAATTATACAGAATGTTGGCCCATGAGGATTTAAGGAAAGCTGCAGTCCTTATCTTTGCAAATAAACAGGATATGAAAGGGTGCATGACAGCAGCTGAAATCTCCAAATACCTCACCCTAAGTTCAATCAAGGACCACCCGTGGCACATCCAGTCGTGCTGTGCGCTCACAGGAGAAGGGTTGTGCCAAGGTCTAGAGTGGATGACCTCCAGGATTGGTGTGAGATAATTTTTTTGCTTGAAAAAGACTTCTCTATTTATTCGGTGATATGAACATTTTTCCTAATACCTTTGGCTGCTGAGGCAGCAGCATGTTTAATTTATGACAACACAAACCTCCAAGTGACGCT
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... ARL5B(75869), Arl5b(75869)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | ||
|---|---|---|
| description targeting mouse Arl5b | description targeting human ARL5B | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| product line MISSION® | product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies |
| Gene Information mouse ... ARL5B(75869), Arl5b(75869) | Gene Information human ... ARL5B(221079) | Gene Information human ... NSUN6(221078) |
| esiRNA cDNA target sequence GTGGGATATCGGTGGTCAAGAATCTCTGAGGTCATCCTGGAACACCTATTACTCAAACACAGAGTTCATCATTCTTGTTGTGGATAGCATTGACAGGGAACGCCTGGCTATTACGAAAGAAGAATTATACAGAATGTTGGCCCATGAGGATTTAAGGAAAGCTGCAGTCCTTATCTTTGCAAATAAACAGGATATGAAAGGGTGCATGACAGCAGCTGAAATCTCCAAATACCTCACCCTAAGTTCAATCAAGGACCACCCGTGGCACATCCAGTCGTGCTGTGCGCTCACAGGAGAAGGGTTGTGCCAAGGTCTAGAGTGGATGACCTCCAGGATTGGTGTGAGATAATTTTTTTGCTTGAAAAAGACTTCTCTATTTATTCGGTGATATGAACATTTTTCCTAATACCTTTGGCTGCTGAGGCAGCAGCATGTTTAATTTATGACAACACAAACCTCCAAGTGACGCT | esiRNA cDNA target sequence GTGGGATATTGGTGGTCAGGAGTCTCTGCGATCATCCTGGAACACATATTACTCAAATACAGAGTTCATCATTCTTGTTGTTGATAGCATTGACAGGGAACGACTAGCTATTACAAAAGAAGAATTATACAGAATGTTGGCTCATGAGGATTTACGGAAGGCTGCAGTCCTTATCTTTGCAAATAAACAGGATATGAAAGGGTGTATGACAGCAGCTGAAATCTCGAAATACCTCACCCTTAGTTCAATTAAGGATCATCCATGGCACATTCAATCCTGCTGTGCTCTCACAGGAGAAGGGTTATGCCAAGGTCTAGAGTGGATGACCTCCCGGATTGGTGTGAGATAACTTTTTTGCTTGAAAGAGACTGCTCTATTTATTCTGTGACATGAACATTTTTTCCTAGTACCTTTGGCTGCTAAGGCAGCAGCATGTTT | esiRNA cDNA target sequence - |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. - | Ensembl | mouse accession no. - |
| form lyophilized powder | form lyophilized powder | form buffered aqueous glycerol solution |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

