Skip to Content
Merck

SML2276

TH-263

≥98% (HPLC)

Synonym(s):

4-[(Phenylamino)sulfonyl]-N-(phenylmethyl)benzamide, N-Benzyl-4-(benzylsulfamoyl)benzamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H18N2O3S
CAS Number:
Molecular Weight:
366.43
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C=C1)S(NC2=CC=CC=C2)(=O)=O)NCC3=CC=CC=C3

Biochem/physiol Actions

Inactive analog control for the type III allosteric LIM-kinase (LIMK) inhibitors TH-251, TH-255 and TH-257.
TH263 is an inactive analog control for the type III allosteric LIM-kinase (LIMK) inhibitors TH-257, TH-255 and TH-251. For characterization details of the active probe, TH-257, please visit the TH-257 probe summary on the Structural Genomics Consortium (SGC) website.

TH-257, the active probe, is available from Sigma. To learn more about and purchase TH-257, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

TH-236 is a negative control for TH-257, a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC chemical probes, visit sigma.com/SGC


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library