Merck
  • Mitochondrial myopathy with dystrophic features due to a novel mutation in the MTTM gene.
Mitochondrial myopathy with dystrophic features due to a novel mutation in the MTTM gene.
Muscle & nerve (2014-04-09)
Lorenzo Peverelli, Carl A Gold, Ali B Naini, Kurenai Tanji, H Orhan Akman, Michio Hirano, Salvatore Dimauro
ABSTRACT
A 61-year-old woman with a 5-year history of progressive muscle weakness and atrophy had a muscle biopsy characterized by a combination of dystrophic features (necrotic fibers and endomysial fibrosis) and mitochondrial alterations [ragged-red, cytochrome c oxidase (COX)-negative fibers]. Sequencing of the whole mtDNA, assessment of the mutation load in muscle and accessible nonmuscle tissues, and single fiber polymerase chain reaction. Muscle mitochondrial DNA (mtDNA) sequencing revealed a novel heteroplasmic mutation (m.4403G>A) in the gene (MTTM) that encodes tRNA(Met). The mutation was not present in accessible nonmuscle tissues from the patient or 2 asymptomatic sisters. The clinical features and muscle morphology in this patient are very similar to those described in a previous patient with a different mutation, also in MTTM, which suggests that mutations in this gene confer a distinctive "dystrophic" quality. This may be a diagnostic clue in patients with isolated mitochondrial myopathy.
Product Description
Supelco
Acrylamide solution, 40% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Supelco
Acrylamide, analytical standard
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Acrylamide, purum, ≥98.0% (GC)
Sigma-Aldrich
Acrylamide, for Northern and Southern blotting, powder blend
Supelco
Acrylamide, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid