Merck
  • Analysis of trihalomethane precursor removal from sub-tropical reservoir waters by a magnetic ion exchange resin using a combined method of chloride concentration variation and surrogate organic molecules.
Analysis of trihalomethane precursor removal from sub-tropical reservoir waters by a magnetic ion exchange resin using a combined method of chloride concentration variation and surrogate organic molecules.
The Science of the total environment (2015-09-12)
Athit Phetrak, Jenyuk Lohwacharin, Satoshi Takizawa
ABSTRACT
In small reservoirs in tropical islands in Japan, the disinfection by-product formation potential is high due to elevated concentrations of dissolved organic matter (DOM) and bromide. We employed a combined method of variation of chloride concentrations and the use of DOM surrogates to investigate removal mechanisms of bromide and different fractions of DOM by chloride-based magnetic ion exchange (MIEX®) resin. The DOM in reservoir waters was fractionated by resins based on their hydrophobicity, and characterized by size-exclusion chromatography and fluorescence excitation-emission matrix spectrophotometry. The hydrophobic acid (HPO acid) fraction was found to be the largest contributor of the trihalomethane (THM) precursors, while hydrophilic acid (HPI acid) was the most reactive precursors of all the four THM species. Bromide and DOM with a molecular weight (MW) greater than 1kDa, representing HPO acid (MW 1-3kDa) and HPI acid (MW 1-2kDa), were effectively removed by MIEX® resin; however, DOM with a MW lower than 1kDa, representing HPI non-acid, was only moderately removed. The removal of THM precursors by MIEX® resin was interfered by high chloride concentrations, which was similar to the removal of glutamic acid (HPI acid surrogate) and bromide. However, elevated chloride concentrations had only a minor effect on tannic acid (HPO acid surrogate) removal, indicating that HPO acid fraction was removed by a combination of ion exchange and physical adsorption on MIEX® resin. Our study demonstrated that the combined use of DOM surrogates and elevated chloride concentrations is an effective method to estimate the removal mechanisms of various DOM fractions by MIEX® resin.
Sigma-Aldrich
Potassium bromide, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium hydroxide solution, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium hydroxide, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Sodium hydroxide solution, 50% in H2O
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, flakes
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium hydroxide, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Sodium hydroxide, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
Sodium hydroxide, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium hydroxide solution, 5.0 M
Sigma-Aldrich
Sodium hydroxide, reagent grade, 97%, powder
Sigma-Aldrich
Sodium hydroxide, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture