Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41122107
eCl@ss:
32049090
Material:
PC membrane
Pore size:
8 μm
Sterility:
sterile
Binding type:
non-treated surface
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
PC membrane
description
cellGrade™ plus
sterility
sterile
packaging
pack of 12 ea (strips of 4 inserts, individually wrapped)
manufacturer/tradename
BRAND 782861
size
6 well
pore size
8 μm
binding type
non-treated surface
General description
Innovative cell culture plates and inserts for manual and automatic cell and tissue culture. The BRANDplates® Insert System, developed in conjunction with the Fraunhofer company, offers an innovative expansion to BRAND′s existing cell culture product range. It consists of two different cell culture plates and two designs of cell culture inserts that can be combined in different configurations.Insert specification:4x insert strips, can be divided into up to 4 individual inserts - hanging position in wellHanging inserts – distance between the well base and insert membrane is 4 mmIndividually packaged strips inserted in thermoformed beakers with 6 strips each in the 6-well cell culture platecellGrade™ plus (hydrophilic cell culture-) surface.Smooth wall inserts: For standard applications like secretion studies, co-cultivations, chemotaxis tests, etc.PS cellGrade™ plus surface, sterile, strip of 4 inserts (divisible).BIO-CERT® CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, Pyrogens and non cytotoxic
Legal Information
BIO-CERT is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
BRANDplates is a registered trademark of BRAND GMBH + CO KG
cellGrade is a trademark of BRAND GMBH + CO KG
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Inlet Opening System, PC membrane, pore size 8 μm | description Inlet Opening System, PC membrane, pore size 1 μm | description PC membrane, pore size 8 μm | description Inlet Opening System, PET membrane, pore size 0.4 μm |
| binding type non-treated surface | binding type - | binding type non-treated surface | binding type - |
| pore size 8 μm | pore size 1 μm | pore size 8 μm | pore size 0.4 μm |
| sterility sterile | sterility sterile | sterility sterile | sterility sterile |
| manufacturer/tradename BRAND 782861 | manufacturer/tradename BRAND 782821 | manufacturer/tradename BRAND 782860 | manufacturer/tradename BRAND 782811 |
| material PC membrane | material PC membrane | material PC membrane | material PET membrane |
| size 6 well | size 6 well | size 6 well | size 6 well |
Still not finding the right product?
Explore all of our products under BRAND® insert strips for 24 x 6-well BRANDplates®
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?