Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.03
UNSPSC Code:
42142600
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócmaterial
Microliter syringe
feature
autoclavable, model 701 RN, no needle
manufacturer/tradename
Hamilton
parameter
10-115 °C temp. range (50-239 °F), 2000 psig pressure
I.D. × O.D.
0.485 mm × 6.60 mm
volume
10 μL
General description
The needles on Thin Layer Chromatography (TLC) syringes are coated from the point for 3/4” with a PTFE-coating. The coating reduces the surface tension between the needle and the liquid making it ideal for reproducible sample spotting. The TLC syringe is available as a cemented needle (SNTLC) or as a replacement needle to be installed into a standard removable needle (RN) syringe. To find the appropriate no needle syringe select general syringes and filter by termination and gauge. Choose the “RN” and “No Needle Included” options to see all syringes that can receive TLC needles. To find the appropriate TLC needle select the TLC option under “Needles” in the main product menu.
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Still not finding the right product?
Explore all of our products under Hamilton® TLC syringe
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?