Merck
Z341908
IKA® universal attachments for KS 130/260 basic and control shakers
AS 260.1 universal attachment
Synonim(y):
IKA 0008017400, adjustable tube holder, tube holder, tube holding attachment, tube rack
Informacje o tej pozycji
NACRES:
NB.73
UNSPSC Code:
41103811
Manufacturer/tradename:
IKA
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 1 ea
manufacturer/tradename
IKA
availability
available only in EU
General description
Note: This attachment is for all kinds of vessels; infinitely variable clamping rolls allow universal adaptation to the vessels.
Legal Information
IKA is a registered trademark of IKA-Werke GmbH & Co. KG
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)