Przejdź do
Z678317
Finnpipette® F2 variable volume pipette
volume 5-50 μL , Micro-tip
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1960,00 zł
Informacje o tej pozycji
NACRES:
NB.01
UNSPSC Code:
41121500
Volume:
5-50 μL , Micro-tip
Packaging:
pack of 1 ea
Feature:
color-coded turquoise, electronic: no, single-channel
Manufacturer/tradename:
Finn 4642120
1960,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
​
Pomoc techniczna
Pozwól nam pomóc
feature
color-coded turquoise, electronic: no, single-channel
packaging
pack of 1 ea
manufacturer/tradename
Finn 4642120
volume
5-50 μL , Micro-tip
General description
Finnpipette F2 has a light pipetting action, is easy to use and is fully autoclavable. Accuracy and precision are the most vital aspect of any pipette so the Finnpipette F2 was developed with Advanced Volume Gearing (AVG). This is a self-supporting modular volume adjustment mechanism separated from the pipette body. This improves accuracy and precision by preventing possible effects of hand warmth on the accuracy of measurements.
  • Advanced Volume Gearing mechanism (AVG) ensures increased accuracy and precision
  • Big display with white numbers on black background for increased visibility
  • Double action pipetting button with very light and smooth pipetting action
  • Fully autoclavable
  • Super blow-out piston with volumes of 50μl and below
Legal Information
Finnpipette is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
Z678287Z678376Z678295
description
volume 5-50 μL , Micro-tip
description
volume 0.5-5 μL , Micro-tip
description
volume 1-10 mL
description
volume 1-10 μL , Micro-tip
feature
single-channel, color-coded turquoise, electronic: no
feature
electronic: no, single-channel, color-coded pink
feature
electronic: no, color-coded red, single-channel
feature
color-coded pink, single-channel, electronic: no
manufacturer/tradename
Finn 4642120
manufacturer/tradename
Finn 4642020
manufacturer/tradename
Finn 4642110
manufacturer/tradename
Finn 4642030
volume
5-50 μL , Micro-tip
volume
0.5-5 μL , Micro-tip
volume
1-10 mL
volume
1-10 μL , Micro-tip
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
packaging
pack of 1 ea
Still not finding the right product?
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)