Merck
539624
Pseudosubstratowy inhibitor PKCζ, mirystylowany - Calbiochem
Synonim(y):
Myr-SIYRRGARRWRKL-OH, Protein Kinase Cζ Pseudosubstrate Inhibitor, Myristoylated, PKCζ Pseudosubstrate Inhibitor, Myristoylated - Calbiochem
Informacje o tej pozycji
Wzór liniowy:
C90H154N30O17
NACRES:
NA.28
UNSPSC Code:
12352200
Form:
lyophilized
Storage condition:
OK to freeze
Pomoc techniczna
Pozwól nam pomóc
form
lyophilized
Quality Segment
storage condition
OK to freeze
solubility
soluble (H2O (2 mg/ml))
storage temp.
−20°C
General description
A cell-permeable, reversible, substrate competitive myristoylated form of protein kinase Cζ pseudo-substrate peptide (Cat. No. 539610) that includes amino acids 113-125 of the pseudosubstrate region. Useful for inhibition studies of PKCζ in intact cells. Reported to inhibit insulin-stimulated glucose transport in intact adipocytes (EC50 = 10-20 µM).
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)