Przejdź do
69739
pET-20b(+) DNA - Novagen
Novagen′s pET-20b(+) vector carries an N-terminal pelB signal sequence for potential periplasmic localization
Synonim(y):
wektor, wektor do klonowania, wektor ekspresji
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
10 μg
Sprawdź dostępność w koszyku
1420,00 zł
Informacje o tej pozycji
NACRES:
NA.51
UNSPSC Code:
41106610
eCl@ss:
32150319
1420,00 zł
Sprawdź dostępność w koszyku
​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Pozwól nam pomóc
Quality Segment
technique(s)
molecular cloning: suitable
Disclaimer
This product is sold for internal research use only. Any commercial use of this product, its components, and/or any derivatives thereof (including but not limited to proteins produced using the product or its components) (together and hereinafter the “EMD Product”) requires signature of a written commercial use agreement with EMD Millipore Corporation or its successor-in-interest. Commercial use shall include but not be limited to: (1) use of the EMD Product to manufacture products for sale to third parties; (2) use of the EMD Product to provide services, information, or data to third parties in exchange for consideration; (3) use of the EMD Product for therapeutic, diagnostic or prophylactic purposes (including as part of a device, chip, assay or other product); or (4) resale of the EMD Product, whether or not such EMD Product is resold for research use. Nothing contained herein shall be deemed to represent or warrant that additional third party rights are not required for use of the EMD Product. Please direct any questions on these use restrictions to: licensing@milliporesigma.com.
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
699526974169740
description
Novagen′s pET-20b(+) vector carries an N-terminal pelB signal sequence for potential periplasmic localization
description
Novagen′s pET-31b(+) vector is designed for cloning and high-level expression of peptide sequences fused with the 125aa ketosteroid isomerase protein.
description
Novagen′s pET-21a-d(+) vectors carry an N-terminal T7-Tag sequence plus an optional C-terminal His-Tag sequence.
description
Novagen′s pET-21a-d(+) vectors carry an N-terminal T7-Tag sequence plus an optional C-terminal His-Tag sequence.
technique(s)
molecular cloning: suitable
technique(s)
molecular cloning: suitable
technique(s)
molecular cloning: suitable
technique(s)
molecular cloning: suitable
Quality Level
100
Quality Level
100
Quality Level
100
Quality Level
100
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)