Przejdź do zawartości
Merck

Przejdź do

ABE1463

Anti-TopBP1 Antibody

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
100 μL

Przewidywany termin wysyłki21 października 2026zKuehne + Nagel Sp. z o.o.

1880,00 zł

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Conjugate:
unconjugated
Clone:
polyclonal
Application:
ICC, WB
Citations:
-

1880,00 zł


Przewidywany termin wysyłki21 października 2026Szczegóły


​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


conjugate

unconjugated

Quality Segment

antibody form

serum

clone

polyclonal

purified by

(unpurified)

species reactivity

mouse, rabbit, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

immunocytochemistry: suitable, western blot: suitable

UniProt accession no.

target post-translational modification

unmodified

General description

Recombinant protein corresponding to human TopBP1.
~180 kDa observed

Application

Western Blotting Analysis: A representative lot detected TopBP1 in U2OS and Saos-2 (Jeon, Y., et al. (2007). J. Biol. Chem. 282(20):14882-14890).
Western Blotting Analysis: A representative lot detected TopBP1 in mouse embryonic stem cells, early mouse embryos, and 3T3 cells (Jeon, Y., et al. (2011). J. Biol. Chem. 286(7):5414-5422).
Immunocytochemistry Analysis: A representative lot detected TopBP1 in 3T3, MEF, HS68, and IMR90 cells (Jeon, Y., et al. (2011). J. Biol. Chem. 286(7):5414-5422).

Physical form

Rabbit polyclonal serum with 0.05% sodium azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Western Blotting in Saos-2 cell lysate.

Western Blotting Analysis: A 1:5,000 dilution of this antibody detected TopBP1 in 10 µg of Saos-2 cell lysate.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
HPA036738HPA036739HPA064702
description

-

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody

clone

polyclonal

clone

polyclonal

clone

polyclonal

clone

polyclonal

antibody form

serum

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

conjugate

unconjugated

species reactivity

mouse, rabbit, human

species reactivity

human

species reactivity

human

species reactivity

human

UniProt accession no.

Q92547

UniProt accession no.

Q92547

UniProt accession no.

Q92547

UniProt accession no.

O00178

technique(s)

immunocytochemistry: suitable, western blot: suitable

technique(s)

immunofluorescence: 0.25-2 μg/mL

technique(s)

immunofluorescence: 0.25-2 μg/mL

technique(s)

immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:1000-1:2500


Klasa składowania

12 - Non Combustible Liquids

Co się dzieje?

WGK 1

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

Nie widzisz odpowiedniej wersji?

Jeśli potrzebujesz konkretnej wersji, możesz wyszukać konkretny certyfikat według numeru partii lub serii.

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów