Przejdź do zawartości
Merck

Przejdź do

ABE566

Anti-phospho Histone H3 (Tyr41) Antibody| ABE566

from rabbit, purified by affinity chromatography

Synonim(y):

Histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
100 μg

Przewidywany termin wysyłki10 grudnia 2026zKuehne + Nagel Sp. z o.o.

2000,00 zł

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.41
eCl@ss:
32160702
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
DB, ICC, WB
Citations:
-

2000,00 zł


Przewidywany termin wysyłki10 grudnia 2026Szczegóły


​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

purified by

affinity chromatography

species reactivity

human, mouse

technique(s)

dot blot: suitable, immunocytochemistry: suitable, western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

phosphorylation (pTyr41)

Gene Information

human ... HIST1H3F(8968)

General description

Histones are highly conserved proteins that serve as the structural scaffold for the organization of nuclear DNA into chromatin. The four core histones, H2A, H2B, H3, and H4, assemble into an octamer (2 molecules of each). Subsequently, 146 base pairs of DNA are wrapped around the octamer, forming a nucleosome, the basic subunit of chromatin. Histones are modified post-translationally by the actions of enzymes in both the nucleus and cytoplasm. These modifications regulate DNA transcription, repair, recombination, and replication. The most commonly studied modifications are acetylation, phosphorylation, methylation, and ubiquitination. These modifications can alter local chromatin architecture, or recruit trans-acting factors that recognize specific histone modifications (the "histone code" hypothesis). The modifications occur predominantly on the N-terminal and C-terminal tails that extend beyond the nucleosome core particle.
~17 kDa observed. Uncharacterized band(s) may be observed in some cell lysates.

Immunogen

Epitope: Near N-terminus
KLH-conjugated linear peptide corresponding to a region near the N-terminus of human Histone H3 phosphorylated at Tyr41.

Application

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho Histone H3 (Tyr41) in HeLa and NIH/3T3 cells.

DyLight® is a registered trademark of Thermo Fisher Scientific.

Dot Blot Analysis: 0.5 µg/mL from a representative lot detected phospho Histone H3 (Tyr41) in an Absurance Histone H3 Antibody Specificity Array (Cat. No. 16-667) and an Absurance Histone H2A, H2B, H4 Antibody Specificity Array (Cat. No. 16-665).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Histones
This Anti-phospho Histone H3 (Tyr41) Antibody is validated for use in western blotting, ICC & dot blot for the detection of phospho Histone H3 (Tyr41).

Physical form

Affinity purified
Purified rabbit polyclonal in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Evaluated by Western Blotting in colcemid treated HeLa acid extract.

Western Blotting Analysis: 1 µg/mL of this antibody detected phospho Histone H3 (Tyr41) in 10 µg of colcemid treated HeLa acid extract.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

DyLight is a registered trademark of Pierce Biotechnology, Inc.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
ABE1057ABE154ABE460
description

from rabbit, purified by affinity chromatography

description

from rabbit, purified by affinity chromatography

description

from rabbit, purified by affinity chromatography

description

serum, from rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

serum

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

Gene Information

human ... HIST1H3F(8968)

Gene Information

human ... HIST1H3F(8968)

Gene Information

human ... HIST1H3F(8968)

Gene Information

human ... HIST1H3F(8968)

technique(s)

immunocytochemistry: suitable, dot blot: suitable, western blot: suitable

technique(s)

immunocytochemistry: suitable, dot blot: suitable, western blot: suitable

technique(s)

western blot: suitable, ChIP: suitable (ChIP-seq)

technique(s)

dot blot: suitable, western blot: suitable, immunohistochemistry: suitable, ChIP: suitable


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

12 - Non Combustible Liquids

Co się dzieje?

WGK 1

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów




Questions

Reviews

No rating value

Active Filters