Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Przewidywany termin wysyłki09 października 2026zKuehne + Nagel Sp. z o.o. | 1870,00 zł |
Informacje o tej pozycji
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7010, for use with Milli-Q® IQ 7015
General description
The valve stops feed water to Milli-Q® IQ 7003/05/10/15 or Milli-Q® IX 7003/05/10/15 water purification systems.
Application
Features and Benefits
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Regulates feed water flow, for use with Milli-Q® IQ/IX systems | description For use with Milli-Q® CLX / HR / HX systems | description Aseptic sampling valve, for use with Milli-Q® IQ/IX systems | description Safety accessory that detects a water leak, for use with Milli-Q® IQ/IX/EQ/SQ series. |
| compatibility for use with Milli-Q® IQ 7015, for use with Milli-Q® IQ 7005, for use with Milli-Q® IX 7005, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7003, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7010 | compatibility for use with Milli-Q® HR, for use with Milli-Q® CLX, for use with Milli-Q® HX | compatibility for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7003, for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7003, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7005, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7015 | compatibility for use with Milli-Q® IX 7010, for use with Milli-Q® IX 7015, for use with Milli-Q® IQ 7000, for use with Milli-Q® IX 7003, for use with Milli-Q® SQ 2Series, for use with Milli-Q® IQ 7005, for use with Milli-Q® IQ 7003, for use with Milli-Q® IQ 7015, for use with Milli-Q® EQ 7016, for use with Milli-Q® EQ 7008, for use with Milli-Q® IQ 7010, for use with Milli-Q® IX 7005, for use with Milli-Q® EQ 7000 |
| packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 unit |
Still not finding the right product?
Explore all of our products under External Solenoid valve
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?