Przejdź do zawartości
Merck

Przejdź do

MABF3286

Anti-CD4 Antibody, clone SK3

Synonim(y):

T-cell surface antigen T4/Leu-3, T-cell surface glycoprotein CD4

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
SK3, monoclonal
Species reactivity:
human
Application:
FACS, IHC
Citations:
-
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

SK3, monoclonal

form

liquid

mol wt

calculated mol wt 51.11 kDa, observed mol wt ~N/A kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100

technique(s)

flow cytometry: suitable, immunohistochemistry: suitable

isotype

IgG1κ

epitope sequence

Extracellular domain

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... CD4(920)

General description

T-cell surface glycoprotein CD4 (UniProt: P01730; also known as T-cell surface antigen T4/Leu-3, CD4) is encoded by the CD4 gene (Gene ID: 920) in human. CD4 is a single-pass type I membrane glycoprotein that is highly expressed in T-helper cells and is also found in macrophages, dendritic cells, and NK cells. CD4 is synthesized with a signal peptide (a 1-25), which is subsequently cleaved off to generate the mature form that has an extracellular domain (aa 26-396), a transmembrane domain (aa 397-418), and a cytoplasmic domain (aa 419-458). It has three Ig-like C2-type and one Ig-like V-type domain. CD4 plays an essential role in the immune response and serves multiple functions in response against both external and internal offenses. In T-cells, it functions primarily as a coreceptor for MHC class II molecule:peptide complex. It interacts simultaneously with the T-cell receptor (TCR) and the MHC class II presented by antigen presenting cells and recruits the Src kinase LCK to the vicinity of the TCR-CD3 complex. The LCK can then initiate different intracellular signaling events by phosphorylating various substrates, which lead to lymphokine production, motility, adhesion and activation of T-helper cells. CD4 is also reported to participates in the development of T-helper cells in the thymus and trigger the differentiation of monocytes into functional mature macrophages. CD4 also serves a primary receptor for human immunodeficiency virus-1 (HIV-1). It is removed from plasma membrane by Nef protein of HIV-1, which increases clathrin-dependent endocytosis and lysosomal degradation of CD4.

Immunogen

Human peripheral blood T lymphocytes.

Application

Quality Control Testing

Evaluated by Flow Cytometry in human peripheral blood mononuclear cells (PBMC).

Flow Cytometry Analysis: 1.0 µg from a representative lot detected CD4/Leu3 in human peripheral blood mononuclear cells (PBMC).

Tested Applications

Immunohistochemistry Applications: A representative lot detected CD4 in Immunohistochemistry applications (Tjin, E.P.M., et al. (2014). Cancer Immunol Res. 2(6):538-46).

Flow Cytometry Analysis: A representative lot detected CD4 in Flow Cytometry applications (Evans, R.L., et al. (1981). Proc Natl Acad Sci USA. 78(1):544-8; Mold, J.E., et al. (2021). Cell Rep. 35(8):109174).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone SK3 is a mouse recombinant monoclonal antibody that detects CD4. It targets an epitope from the extracellular domain.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
MABF172BSAB4503583SAB4700054
description

-

description

clone SK3, from mouse, Peridinin-Chlorophyll-Protein Complex

description

affinity isolated antibody

description

clone MEM-16, purified immunoglobulin, buffered aqueous solution

antibody form

purified antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified immunoglobulin

biological source

mouse

biological source

mouse

biological source

rabbit

biological source

mouse

species reactivity

human

species reactivity

human

species reactivity

human, mouse

species reactivity

human

clone

SK3, monoclonal

clone

SK3, monoclonal

clone

polyclonal

clone

MEM-16, monoclonal

Gene Information

human ... CD4(920)

Gene Information

human ... CD4(920)

Gene Information

human ... CD4(920)

Gene Information

human ... CD4(920)

UniProt accession no.

P01730

UniProt accession no.

P01730

UniProt accession no.

P01730

UniProt accession no.

P01730


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

12 - Non Combustible Liquids

WGK

WGK 1

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów