Przejdź do zawartości
Merck

Przejdź do

MABS2274

Anti-PKC δ Antibody, clone 10F11B

Synonim(y):

EC:2.7.10.2, EC:2.7.11.13, Protein kinase C delta type, Protein kinase C delta type catalytic subunit, Protein kinase C delta type regulatory subunit, Sphingosine-dependent protein kinase-1 (SDK1), Tyrosine-protein kinase PRKCD (EC:2.7.10.2), nPKC-delta

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
25 μg
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
723,00 zł
100 μg
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
1830,00 zł

Informacje o tej pozycji

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
10F11B, monoclonal
Species reactivity:
mouse, human
Application:
WB
Citations:
-

723,00 zł


Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności

​
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


biological source

mouse

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

10F11B, monoclonal

mol wt

calculated mol wt 77.51 kDa, observed mol wt ~77 kDa

purified by

using protein G

species reactivity

mouse, human

packaging

antibody small pack of 100

technique(s)

western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... PRKCD(5580)

General description

Protein kinase C delta type (UniProt: Q05655; also known as EC:2.7.11.13, Tyrosine-protein kinase PRKCD (EC:2.7.10.2), EC:2.7.10.2, nPKC-delta, Protein kinase C delta type regulatory subunit, Protein kinase C delta type catalytic subunit, Sphingosine-dependent protein kinase-1 (SDK1)) is encoded by the PRKCD (also known as PKCD) gene (Gene ID: 5580) in human. Protein kinase C delta type (PKC ) is a serine/threonine kinase, a member of the novel protein kinase C (PKC) subfamily. PKC comprises several domains including a regulatory domain with C1 and C2 motifs that mediate its interaction with diacylglycerol and phospholipids, and a catalytic domain responsible for its kinase activity. PKC is involved in regulating diverse cellular processes such as apoptosis, cell proliferation, differentiation, growth regulation, tissue remodeling and immune responses. It acts as a signal transducer in pathways triggered by various stimuli, including growth factors and stress signals, by phosphorylating a wide range of substrates. PKC is ubiquitously expressed, with particularly high levels in tissues such as the brain, heart, and immune cells, reflecting its broad functional roles. PKC has been implicated in cancer, where it can act as either a tumor suppressor or promoter depending on the cellular context. It is also associated with cardiovascular diseases, diabetes, and autoimmune disorders, and Parkinson where its dysregulation can contribute to disease pathogenesis. (Ref.: Miao. L., et al., (2022) Front. Cardiovasc. Med. 9, Article 816369)

Immunogen

Full-length, recombinant human PKC .

Application

Quality Control Testing

Evaluated by Western Blotting in RAW 264.7 cell lysate.

Western Blotting Analysis: A 1:250 dilution from a representative lot detected PKC in RAW 264.7 cells lysate.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Biochem/physiol Actions

Clone 10F11B is a mouse recombinant monoclonal antibody that detects PKC .

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Preparation Note

0.5 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.
Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
SAB4502360ZRB1623SAB5700371
description

-

description

affinity isolated antibody

description

recombinant, expressed in HEK 293 cells

description

-

species reactivity

mouse, human

species reactivity

human, mouse, rat

species reactivity

rat, mouse, human

species reactivity

mouse, human

biological source

mouse

biological source

rabbit

biological source

rabbit

biological source

rabbit

clone

10F11B, monoclonal

clone

polyclonal

clone

1C13, monoclonal, recombinant monoclonal

clone

polyclonal

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

Gene Information

human ... PRKCD(5580)

Gene Information

-

Gene Information

-

Gene Information

-

UniProt accession no.

Q05655

UniProt accession no.

Q05655

UniProt accession no.

-

UniProt accession no.

Q05655


Still not finding the right product?

Wypróbuj nasze narzędzie Narzędzie selektora produktów, aby zawęzić opcje.


Klasa składowania

12 - Non Combustible Liquids

WGK

WGK 3

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów