Przejdź do
MXQUA4VP2
Milliflex® Quantum system Validation Protocol for the combination with Milliflex® Oasis system
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 unit
Dostępne do wysyłki DZISIAJzKuehne + Nagel Sp. z o.o.
16 170,00 zł
Informacje o tej pozycji
NACRES:
ND.05
UNSPSC Code:
41116500
eCl@ss:
32150319
16 170,00 zł
Dostępne do wysyłki DZISIAJSzczegóły
​
Więcej informacji
Pomoc techniczna
Pozwól nam pomóc
availability
not available in North America
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
General description
Milliflex® Quantum Validation Protocol(A4 Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of rapid microbial detection system using Milliflex® Quantum in combination with Milliflex® Oasis system. The protocol contains an exhaustive list of tests including test descriptions and test records. The protocol follows GMP, EP, USP and JP regulatory guidelines. Not available in North America.
Application
Milliflex® Quantum Validation Protocol is used to validate your rapid microbial detection system with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
MXQULTVP1MXQUA4VP1MM2PA4VP1
description
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
description
The Milliflex Quantum validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications.
description
The Milliflex Quantum validation protocol is available to make validation faster and easier and ensures that your system′s performance remains compliant with its validated specifications.
description
-
compatibility
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
compatibility
-
compatibility
-
compatibility
for use with MILLIFLEX®
availability
not available in North America
availability
-
availability
-
availability
-
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)