Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Przewidywany termin wysyłki30 listopada 2026zKuehne + Nagel Sp. z o.o. | 2390,00 zł |
Informacje o tej pozycji
packaging
pkg of 1 unit
manufacturer/tradename
Milliflex®
technique(s)
Rapid Sterility Testing: suitable
application(s)
bioburden testing
pharmaceutical, sterility testing
compatibility
for use with Milliflex® Rapid System 2.0
shipped in
ambient
General description
Application
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description The Milliflex® Rapid2.0 detection tower is suitable for bioburden and sterility testing, suitable for sterility testing | description - | description The Milliflex® Rapid2.0 detection tower is suitable for bioburden and sterility testing, for use with Milliflex® Rapid System 2.0, suitable for sterility testing | description Milliflex Oasis pumppower supply for WEU, for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station |
| manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® | manufacturer/tradename Milliflex® |
| packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 unit | packaging pkg of 1 ea |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in - |
| application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing | application(s) bioburden testing |
| compatibility for use with Milliflex® Rapid System 2.0 | compatibility for use with Milliflex® Rapid System 2.0 | compatibility for use with Milliflex® Rapid System 2.0 | compatibility for use with the Milliflex Rapid 2.0 Detection Tower or the Milliflex Rapid 2.0 Autospray Station |
| technique(s) Rapid Sterility Testing: suitable | technique(s) Rapid Sterility Testing: suitable | technique(s) Rapid Sterility Testing: suitable | technique(s) Rapid Sterility Testing: suitable |
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?