Przejdź do
RDYSTRMPUMP
ReadyStream® Pump
Supplies water from an external pump, for use with ReadyStream® system
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
Przewidywany termin wysyłki12 listopada 2026zMerck Life Science sp. z o.o.
6180,00 zł
Informacje o tej pozycji
UNSPSC Code:
41116107
NACRES:
NA.71
6180,00 zł
Przewidywany termin wysyłki12 listopada 2026Szczegóły
​
Więcej informacji
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc
Quality Segment
agency
ISO 11133
feature
Supplies water from an external pump
manufacturer/tradename
ReadyStream®
application(s)
food and beverages
compatibility
for use with ReadyStream® system
General description
The ReadyStream® Pumpis engineered to efficiently supply water that complies with ISO 11133 from anexternal tank to the ReadyStream® system. It features quick connector tubingfor seamless integration with the ReadyStream® system, ensuring a hassle-freesetup. The pump is compatible with Milli-Q tanks when used with the dedicatedconnection kit (RDYSTRMMQ01). Designed for optimal performance, the pumpautomatically starts and stops to maintain the required pressure and flow ratenecessary for the effective operation of the ReadyStream® system, making it areliable solution
Application
The ReadyStream® Pumpis primarily used in laboratory and industrial settings where precise waterquality is crucial for various processes. It is ideal for feeding water to the ReadyStream® system, ensuring compliance with stringent quality standardsoutlined in ISO 11133.
Legal Information
READYSTREAM is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
RDYSTRM20RDYSTRM01EURDYSTRM10
description
Supplies water from an external pump, for use with ReadyStream® system
description
For convenient dispensing of large amount of food testing media in short time
description
For convenient preparation of large amounts of food testing media in a short time
description
For convenient preparation of large amount of food testing media in short time
application(s)
food and beverages
application(s)
food and beverages
application(s)
food and beverages
application(s)
food and beverages
Quality Level
EQ2
Quality Level
EQ2
Quality Level
EQ2
Quality Level
EQ2
feature
Supplies water from an external pump
feature
-
feature
-
feature
-
manufacturer/tradename
ReadyStream®
manufacturer/tradename
ReadyStream®
manufacturer/tradename
-
manufacturer/tradename
ReadyStream®
agency
ISO 11133
agency
-
agency
ISO 11133
agency
-
compatibility
for use with ReadyStream® system
compatibility
for use with ReadyStream® system
compatibility
-
compatibility
for use with ReadyStream® System
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)