Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.
Wybierz wielkość
Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
UNSPSC Code:
41106514
NACRES:
NA.81
Biological source:
mouse
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomócbiological source
mouse
Quality Segment
packaging
vial of ≥1X10⁶ vial (viable cells per vial)
manufacturer/tradename
Millipore
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
General description
Hematopoietic stem cells (HSCs) are a type of stem cell that differentiate into all other mature blood cells derived from two lineages, myeloid and lymphoid. Hematopoiesis, a process in which HSCs differentiate into other mature blood cell lineages, occurs in the bone marrow, and gives HSCs unique regenerative properties in which they are capable of generating a variety of cell types as well as generating more stem cells in the process. HSCs migrate to injury sites and are useful for tissue repair. HSCs also appear to have potential for therapeutic treatment in inflammatory bowel disease models. HSCs have already displayed some therapeutic benefit experimentally in models used for colitis and Crohn′s disease. Clinical use of HSCs through systemic injection has been quite limited by their rarity and poor tissue retention. Understanding HSC recruitment remains an important topic for improving stem cell-based therapies. HPC-7, a mouse HSC cell line, was developed to further understand hematopoiesis and functionality of HSCs. The HPC-7 cell line retains many cell characteristics typical of fetal-derived hematopoietic precursors such as being c-kit+, CD34+, CD44+, Sca-1-, and Thy1.2-. Transcription factors expressed by the HPC-7 cell line are also consistent with characterized HSCs. HPC-7 cells can be differentiated through the addition of different cell factors and have been shown to change cell-surface expression post-differentiation. HPC-7 cells also retain the self-renewal properties characteristic of HSCs. Source: Mouse LH2 cDNA was retrovirally expressed in embryonic stem cells and subjected to differentiation assay in vitro. This led to the generation of multipotent hematopoietic precursor cells labeled HPC-7.ReferencesPinto do O P. 1998. Expression of the LIM-homeobox gene LH2 generates immortalized Steel factor-dependent multipotent hematopoietic precursors. The EMBO Journal. 17(19):5744–5756.
Application
- Cells are verified to be of mouse origin and negative for human, rat, Chinese hamster, Golden Syrian hamster, and non-human primate interspecies contamination, as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
- Cells tested negative for infectious diseases against a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Cells tested negative for mycoplasma.
Features and Benefits
The HPC-7 cell line retains many cell characteristics of fetal-derived hematopoietic precursors such as being c-kit+, CD34+, CD44+, Sca-1-, and Thy1.2-.
Preparation Note
The cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Other Notes
This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the “Academic Use Agreement” as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Ta strona może zawierać tekst przetłumaczony maszynowo.
1 of 1
Ta pozycja | |
|---|---|
| description - | description Pharmaceutical Secondary Standard; Certified Reference Material |
| technique(s) cell culture | mammalian: suitable | technique(s) gas chromatography (GC): suitable, HPLC: suitable |
| biological source mouse | biological source - |
| shipped in liquid nitrogen | shipped in - |
| storage temp. −196°C | storage temp. 2-30°C |
| manufacturer/tradename Millipore | manufacturer/tradename - |
| Quality Level 100 | Quality Level 300 |
Klasa składowania
12 - Non Combustible Liquids
Co się dzieje?
WGK 2
Temperatura zapłonu (°F)
Not applicable
Temperatura zapłonu (°C)
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

