Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
Quality Segment
biological source
human breast
packaging
vial of ≥1200 viable organoids vial
manufacturer/tradename
Millipore
gender
female
technique(s)
cell culture: suitable
application(s)
3D cell culture
control
breast
relevant disease(s)
carcinoma
shipped in
liquid nitrogen
storage temp.
−196°C
General description
Worldwide, breast cancer (BC) is the most commonly diagnosed cancer among women. In the United States, BC accounts for 1 in 3 of all new female cancer cases each year. BC consists of at least 21 distinct histological subtypes and four major molecular subtypes. The grade and cell proliferation may also be used to classify BC subtypes. Almost 81% BC are invasive, and >75% of such invasive cancers are ductal carcinomas, commonly called invasive ductal carcinoma (IDC). It is important to note that the subtypes show overlaps and there maybe differences in clinical approximations and molecular methods to classify BCThe major molecular subtypes are:
- Luminal A (HR+/HER2-): This subtype expresses hormone receptors (HR) for estrogen, progesterone, or both. This is the most common BC subtype, is less aggressive and responds to hormone therapy.
- Luminal B (HR+/HER2+): This subtype expresses the epidermal growth factor receptor HER2 and may also be highly positive for the cell proliferation marker Ki67. Luminal B subtype usually shows poorer prognosis than Luminal A.
- Triple negative or basal-like (HR-/HER2-): This subtype does not express HR or HER2, and therefore have the poorest outcome since as it is minimally responsive to hormone therapy or HER2-targeted treatment. Triple negative BC is more common in black or premenopausal women and also in patients with BRCA1 mutation.
- HER2-enriched (HR-/HER2+): With recent advances in HER2-targeted therapies (such as cell therapy), this subtype now shows significantly improved prognosis.
Cell Line Characteristics :
Sex: Female
Age: 68
Organ: Breast (Tumor)
Disease: Infiltrating Ductal Carcinoma (IDC), ER+/PR+/HER2-
History: High blood pressure (Almodipine/Spironolactone), cholesterol (Pravastatin), asthma (Atrovent/Singulair), migraine (Rizatriptan/Topiramate), nerve pain (Gabapentin/Tramadol), depression (Zoloft).
3dGRO® PDO were derived utilizing HUB Organoid Technology. The purchaser of this product shall agree to HUB’s terms of use (listed below), which shall be separately acknowledged and accepted by such purchaser, prior to transfer of this product to purchaser.
Application
- Viability: >1500 viable organoids/vial
- Organoid Growth: Pass
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Mycoplasma Contamination: Negative
- STR Profile: Pass
Features and Benefits
Preparation Note
Legal Information
3dGRO® organoids were derived utilizing HUB Organoid Technology.
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
Disclaimer
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Infiltrating Ductal Carcinoma, Breast, Age: 68, Female, Mutation: Not Available | description Infiltrating Ductal Carcinoma, Breast, Age: 84, Female, Mutation: Notch | description Infiltrating Ductal Carcinoma, Breast, Age: 58, Female, Mutation: Not Available | description Adenocarcinoma, Colon, Age: 47, Female, Mutation: Not Available |
| technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable |
| biological source human breast | biological source human breast | biological source human breast | biological source human colon |
| relevant disease(s) carcinoma | relevant disease(s) carcinoma (Infiltrating Ductal) | relevant disease(s) carcinoma (Infiltrating Ductal) | relevant disease(s) adenocardinoma |
| application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture |
| storage temp. −196°C | storage temp. −196°C | storage temp. −196°C | storage temp. −196°C |
| packaging vial of ≥1200 viable organoids vial | packaging vial of ≥1500 vial (viable cells per vial) | packaging vial of ≥1500 vial (viable cells per vial) | packaging vial of ≥1500 organoids/vial vials |
Klasa składowania
12 - Non Combustible Liquids
WGK
WGK 2
Temperatura zapłonu (°F)
No data available
Temperatura zapłonu (°C)
No data available
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.



