Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 vial | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 13 120,00 zł |
Informacje o tej pozycji
13 120,00 zł
Quality Segment
biological source
human liver
packaging
vial of ≥1500 organoids/vial vial
manufacturer/tradename
Millipore
gender
female
technique(s)
cell culture: suitable
application(s)
3D cell culture
control
liver
relevant disease(s)
(normal)
storage temp.
-140 to -196°C
General description
The 3dGRO® human liver organoid biobank is a collection of tissue-derived normal non-diseased human liver organoids from numerous patients. The biobank contains liver PDOs that are highly viable and express high levels of albumin, HNF4-a, FOXA2, SOX17, CK7, CK19, E-Cadherin, PCK2 along with important cytochrome P450 (CYP) enzymes activities (Cyp3A4, Cyp2C9, Cyp2B6, Cyp2D6, Cyp1A2).
Cell Line Characteristics
- Sex: Female
- Age: 51 Years
- Race: Asian (Indian)
- Organ: Liver
- Disease: Normal
- Patient History: None
Application
- Viability: >1500 viable organoids/vial
- Organoid Growth: Pass
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Mycoplasma Contamination: Negative
- STR Profile: Pass
Features and Benefits
Preparation Note
Legal Information
3dGRO® organoids were derived utilizing HUB Organoid Technology.
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact organoids@milliporesigma.com.
Disclaimer
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description Normal, Liver, Age: 51, Female, Mutation: None | description Normal, Liver, Age: 65, Female, Mutation: None | description Normal, Liver, Age: 60, Male, Mutation: None | description Normal, Liver, Age: 60, Female, Mutation: None |
| application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture | application(s) 3D cell culture |
| biological source human liver | biological source human liver | biological source human liver | biological source human liver |
| relevant disease(s) (normal) | relevant disease(s) (normal) | relevant disease(s) (normal) | relevant disease(s) (normal) |
| technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable | technique(s) cell culture: suitable |
| storage temp. -140 to -196°C | storage temp. −196°C | storage temp. -140 to -196°C | storage temp. −196°C |
| gender female | gender female | gender male | gender female |
Klasa składowania
10 - Combustible liquids
Temperatura zapłonu (°F)
Not applicable
Temperatura zapłonu (°C)
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?
Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.



