Przejdź do
ZFC0NDCA1
Feed Water Conductivity Cell
For use with Milli-Q® IQ 7000, To monitor the ionic quality of feed water
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 ea
Przewidywany termin wysyłki28 października 2026zKuehne + Nagel Sp. z o.o.
2560,00 zł
Informacje o tej pozycji
NACRES:
JA.13
UNSPSC Code:
41104200
2560,00 zł
Przewidywany termin wysyłki28 października 2026Szczegóły
​
Więcej informacji
Pomoc techniczna
Pozwól nam pomóc
feature
measures the conductivity of water
packaging
pkg of 1 unit
manufacturer/tradename
Milli-Q®
compatibility
for use with Milli-Q® IQ 7000
General description
The feed water conductivity cell is an installation accessory which triggers an alarm if the feed water quality is out of specifications. This prevents premature exhaustion of IPAK Meta® and IPAK Quanta® cartridges.
Application
The feed water conductivity cell is used to monitor the ionic quality of feed water for Milli-Q® IQ 7000 system.
Legal Information
IPAK META is a registered trademark of Merck KGaA, Darmstadt, Germany
IPAK QUANTA is a registered trademark of Merck KGaA, Darmstadt, Germany
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
ZFC0NDCL1SPKBTP6KINS002ZLXL000PR
description
For use with Milli-Q® IQ 7000, To monitor the ionic quality of feed water
description
for use with Milli-Q® Reference, To monitor the ionic quality of feed water
description
-
description
Regulates the feed water pressure from 0-25 bars, Ensures accurate water level sensing to prevent overflow for use with high flow water systems
compatibility
for use with Milli-Q® IQ 7000
compatibility
for use with Milli-Q® Reference
compatibility
-
compatibility
for use with Milli-Q® CLX
feature
measures the conductivity of water
feature
-
feature
-
feature
-
packaging
pkg of 1 unit
packaging
pkg of 1 unit
packaging
-
packaging
pkg of 1 unit
manufacturer/tradename
Milli-Q®
manufacturer/tradename
Milli-Q®
manufacturer/tradename
-
manufacturer/tradename
-
Still not finding the right product?
Explore all of our products under Feed Water Conductivity Cell
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)