Przejdź do
ZKITRES00SD
Resistivity Kit for High-throughput Systems
For use with Milli-Q® HX 7000 SD water systems
Wybierz wielkość
Zmień widok
Rozmiar/SKUDostępnośćCena netto
1 kit
Przewidywany termin wysyłki12 listopada 2026zKuehne + Nagel Sp. z o.o.
4380,00 zł
Informacje o tej pozycji
NACRES:
JA.13
UNSPSC Code:
41104200
4380,00 zł
Przewidywany termin wysyłki12 listopada 2026Szczegóły
​
Więcej informacji
Pomoc techniczna
Pozwól nam pomóc
packaging
pkg of 1 unit
compatibility
for use with Milli-Q® HX
General description
Installed within the purification system, this resistivity monitor verifies the resistivity level of water in the loop.
Features and Benefits
  • The resistivity booster can be easily installed on the front of the water purification system.
  • The ergonomic and patented pack-locking mechanism allows for rapid replacement when needed.
Legal Information
Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany
Porównaj podobne pozycje
Wyświetl pełne porównanie
Pokaż różnice
1 of 1
Ta pozycja
ZFCBIQ0AUZKITRES01SDZFMQPR3BT0
description
For use with Milli-Q® HX 7000 SD water systems
description
Available only in Australia and New Zealand.
description
For use with Milli-Q® HX 7000 SD water systems
description
Limits tap feed water pressure to 3 bars to ensure optimal RO membrane performance
compatibility
for use with Milli-Q® HX
compatibility
for use with Milli-Q® IQ 7000
compatibility
for use with Milli-Q® HX
compatibility
-
packaging
pkg of 1 unit
packaging
pkg of 1 ea
packaging
pkg of 1 unit
packaging
pkg of 1 unit
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)