Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
0.4 mg | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 2710,00 zł |
Informacje o tej pozycji
2710,00 zł
carrier
PAA carrier
label
FITC labeled
product line
FP-Series
assay
95% (H-NMR)
form
lyophilized powder
mol wt
~20,000 Da
manufacturer/tradename
GlycoNZ
extent of labeling
1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density
technique(s)
flow cytometry: suitable
probe type
disaccharide
probe presentation
polymeric
storage temp.
−20°C
General description
These glycoconjugate probes are designed to reveal lectins, including selectins, directly on cell membranes, organelles, and in cytoplasm. The low fluorescein content ensures that probe solubility in water is not affected, but still allows reliable probe detection. The affinity of the probes is 102 to 105 times higher than that of corresponding free sugars.
PAA is poly[N-(2-hydroxyethyl)acrylamide], a flexible polymer chain which serves as an additional spacer. Used as a molecular weight carrier, PAA demonstrates an absence of non-specific interaction with cells, and is stable to chemical and proteolytic action.
Application
Preparation Note
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description - | description - | description - | description - |
| carrier PAA carrier | carrier PAA carrier | carrier PAA carrier | carrier PAA carrier |
| probe presentation polymeric | probe presentation polymeric | probe presentation polymeric | probe presentation polymeric |
| probe type disaccharide | probe type disaccharide | probe type disaccharide | probe type disaccharide |
| technique(s) flow cytometry: suitable | technique(s) flow cytometry: suitable | technique(s) ELISA: suitable | technique(s) ELISA: suitable |
| product line FP-Series | product line FP-Series | product line PA-Series | product line PA-Series |
| extent of labeling 1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density | extent of labeling 1% Biotin per 20 mole % Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density | extent of labeling 20 mol% Poly[N-(2-hydroxyethyl)acrylamide-co-N-acrylamide] ligand density | extent of labeling - |
Klasa składowania
13 - Non Combustible Solids
WGK
WGK 3
Not applicable
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?