Wybierz wielkość
| Rozmiar/SKU | Dostępność | Cena netto |
|---|---|---|
1 ea | Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności | 464,00 zł |
Informacje o tej pozycji
464,00 zł
material
PTFE tip, glass body
needle size
(not included)
feature
without needle Luer tip
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80601
needle L
, n/a
volume
100 μL
application(s)
general analytical
compatibility
for use with Hamilton 710LT (Luer tip)
General description
Legal Information
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description 710LT, volume 100 μL, needle size (not included) | description 705LT, volume 50 μL, needle size (not included) | description 725LT, volume 250 μL, needle size (not included) | description 750LT, volume 0.5 mL, needle size (not included) |
| material PTFE tip, glass body | material glass body | material glass body | material glass body |
| packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea | packaging pkg of 1 ea |
| volume 100 μL | volume 50 μL | volume 250 μL | volume 0.5 mL |
| manufacturer/tradename Hamilton 80601 | manufacturer/tradename Hamilton 80501 | manufacturer/tradename Hamilton 80701 | manufacturer/tradename Hamilton 80801 |
| needle size (not included) | needle size (not included) | needle size (not included) | needle size (not included) |
| application(s) general analytical | application(s) general analytical | application(s) general analytical | application(s) - |
Still not finding the right product?
Explore all of our products under Hamilton® syringe, luer tip
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?