Przejdź do zawartości
Merck

Przejdź do

PMC2008

AdBrettPhos

≥98%, ISOM8™

Synonim(y):

AdBrettPhos, 2-(Di-1-adamantylphosphino)-3,6-diMethoxy-2′,4′,6′-triisopropyl-1,1′-biphenyl, Di-Ad-BrettPhos

Zaloguj się, aby wyświetlić ceny organizacyjne i kontraktowe.

Wybierz wielkość

Zmień widok
Rozmiar/SKUDostępnośćCena netto
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
2550,00 zł
Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności
8880,00 zł

Informacje o tej pozycji

Wzór empiryczny (zapis Hilla):
C43H61O2P
Numer CAS:
Masa cząsteczkowa:
640.92
MDL number:
UNSPSC Code:
12352128
PubChem Substance ID:
NACRES:
NA.22
eCl@ss:
32151101

2550,00 zł


Skontaktuj się z Obsługą Klienta, aby uzyskać informacje na temat dostępności

​
Złóż zamówienie na produkt na zamówienie
Pomoc techniczna
Potrzebujesz pomocy? Nasz zespół doświadczonych naukowców chętnie Ci pomoże.
Pozwól nam pomóc


product line

ISOM8™

Quality Segment

assay

(HPLC), ≥98%

form

solid

mol wt

640.92

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Heck Reaction, reaction type: Hiyama Coupling, reaction type: Negishi Coupling, reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Suzuki-Miyaura Coupling, reagent type: ligand

mp

231-235 °C

application(s)

diagnostic assay manufacturing

functional group

phosphine

storage temp.

room temp

SMILES string

COc1ccc(OC)c(c1P(C23CC4CC(CC(C4)C2)C3)C56CC7CC(CC(C7)C5)C6)-c8c(cc(cc8C(C)C)C(C)C)C(C)C

InChI

1S/C43H61O2P/c1-25(2)34-17-35(26(3)4)39(36(18-34)27(5)6)40-37(44-7)9-10-38(45-8)41(40)46(42-19-28-11-29(20-42)13-30(12-28)21-42)43-22-31-14-32(23-43)16-33(15-31)24-43/h9-10,17-18,25-33H,11-16,19-24H2,1-8H3

InChI key

NMGHOZQCYNKWBG-UHFFFAOYSA-N

General description

Welcome to our ISOM8™ line of high-quality products available and able to scale for manufacturing. Within our ISOM8 team, we have expertise and documentation to support ISO 9001 industrial and non-GMP pharma production needs, as defined by our M-Clarity™ Program.
Our portfolio of ISOM8 products and chemistries is ever evolving to support our valued Sigma-Aldrich® customers′ requirements. Please navigate to our documents section to look at dossiers and other quality documentation for this product, provided with support of our Elevate Program.

Application

Recent Studies and Innovations:  
  1. Selective Primary Amination of Hindered Arenes: A catalyst system incorporating beta-methylnaphthyl (MeNAP) palladium bromide with AdBrettPhos successfully resolved the competition between single and double arylation, achieving selective introduction of a primary amino group into sterically congested aromatic substrates (Xue et al., 2023).
  2. Amidation of Five-Membered Heterocyclic Bromides: Five-membered heterocycles present significant challenges in palladium-catalyzed amidation due to their electron-rich nature and the propensity of heteroatoms to coordinate and deactivate the catalyst. The use of [{(allyl)PdCl}2] in combination with AdBrettPhos markedly improves catalytic efficiency for these demanding substrates (Su et al., 2012).
  3. Thermally and Air-Stable Precatalysts: Precatalysts derived from PdCl2, Pd(OAc)2, or [(cinnamyl)PdCl]2 in combination with AdBrettPhos exhibit excellent thermal stability at room temperature and notable air/moisture stability, making them ideal for cross-coupling reactions without the need for additional base required by more conventional systems (Lee et al., 2014)

AdBrettPhos is a bulky biarylphosphine ligand developed by the Buchwald group, distinguished by its sterically demanding adamantyl substituents. It enables highly efficient palladium-catalyzed cross-coupling reactions and is particularly noted for forming C-N, C-O, C-F, C-CF3, and C-S bonds under conditions that challenge conventional ligand systems.

Notable Characteristics & Key Uses
  • Sterically Demanding Structure: The bulky adamantyl groups create a highly hindered coordination environment, enabling selective bond formations that are otherwise inaccessible with less congested ligands.
  • Exceptional Stability: AdBrettPhos is frequently deployed as part of air-, moisture-, and thermally stable palladium G3 (3rd Generation) and G6 (6th Generation) precatalysts, simplifying handling while maintaining consistently high catalytic activity.
  • Broad Bond-Forming Capability: Forms a diverse array of bond types - C-C, C-N, C-O, C-F, C-CF3, and C-S - including reactions previously considered difficult or impossible with conventional ligands.
  • Selectivity for Monoarylation: Renowned for its ability to promote selective monoarylation of ammonia and coupling of weak nucleophiles, while effectively suppressing undesirable diarylation side products.
  • User-Friendly Precatalyst Systems: Compatible precatalyst systems allow for low catalyst loadings, abbreviated reaction times, and simplified preparation and handling protocols suitable for research and industrial scales.
  • Suzuki-Miyaura Cross-Coupling (C-C): Enables efficient biaryl and aryl-alkyl coupling across a broad substrate scope, including challenging aryl chlorides.
  • Buchwald-Hartwig Amination (C-N): Particularly powerful for the amidation of challenging five-membered heterocyclic halides and the selective monoarylation of ammonia.
  • Heck, Hiyama, Negishi, Sonogashira, and Stille Couplings: Versatile across multiple palladium-catalyzed C-C bond-forming reactions.
  • C-O Cross-Coupling: Facilitates coupling of primary alcohols with aryl electrophiles to form ethers and related products.
  • Fluorination and Trifluoromethylation: Enables trifluoromethylation of aryl chlorides and synthesis of aryl fluorides, including highly challenging transformations.

Legal Information

ISOM8 is a trademark of Sigma-Aldrich Co. LLC
M-CLARITY is a trademark of Merck KGaA, Darmstadt, Germany
SIGMA-ALDRICH is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives. This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Atom Economy” and “Design for Energy Efficiency”. Click here to view its DOZN scorecard.

Ta strona może zawierać tekst przetłumaczony maszynowo.

Porównaj podobne pozycje

Wyświetl pełne porównanie

Pokaż różnice

1 of 1

Ta pozycja
CDS025458936839768154
description

≥98%, ISOM8™

description

AldrichCPR

description

-

description

95%

reaction suitability

reaction type: Sonogashira Coupling, reaction type: Stille Coupling, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reaction type: Suzuki-Miyaura Coupling, reaction type: Hiyama Coupling, reaction type: Heck Reaction, reaction type: Negishi Coupling, reagent type: ligand

reaction suitability

-

reaction suitability

reagent type: ligand

reaction suitability

reaction type: Negishi Coupling, reaction type: Stille Coupling, reaction type: Hiyama Coupling, reaction type: Buchwald-Hartwig Cross Coupling Reaction, reagent type: ligand, reaction type: Suzuki-Miyaura Coupling, reaction type: Heck Reaction, reaction type: Sonogashira Coupling

assay

(HPLC), ≥98%

assay

-

assay

-

assay

95%

functional group

phosphine

functional group

-

functional group

-

functional group

phosphine

Quality Level

300

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

room temp

storage temp.

-

storage temp.

-

storage temp.

-

form

solid

form

-

form

solid

form

solid


Still not finding the right product?

Explore all of our products under AdBrettPhos


Klasa składowania

11 - Combustible Solids

Temperatura zapłonu (°F)

Not applicable

Temperatura zapłonu (°C)

Not applicable



Wybierz jedną z najnowszych wersji:

Certyfikaty analizy (CoA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Dokumenty section.

Proszę o kontakt, jeśli potrzebna jest pomoc Obsługa Klienta

Masz już ten produkt?

Dokumenty związane z niedawno zakupionymi produktami zostały zamieszczone w Bibliotece dokumentów.

Odwiedź Bibliotekę dokumentów