Merck
AXYPCR48R
Corning® Axygen® PCR Microplate
size 48 wells, conical bottom red polypropylene wells, pkg of 5x10plates/cs
Wybierz wielkość
Informacje o tej pozycji
NACRES:
NB.22
UNSPSC Code:
41121800
Material:
conical bottom red polypropylene wells
Size:
48 wells
Pozwól nam pomóc
Nazwa produktu
Corning® Axygen® PCR Microplate, size 48 wells, conical bottom red polypropylene wells, pkg of 5x10plates/cs
material
conical bottom red polypropylene wells
sterility
non-sterile
manufacturer/tradename
Axygen Inc PCR-48-R
Corning
packaging
pkg of 5x10plates/cs
maximum volume
200 μL
size
48 wells
working volume
200 μL
suitability
suitable for PCR
binding type
non-treated surface
Legal Information
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.
Wybierz jedną z najnowszych wersji:
Certyfikaty analizy (CoA)