Zmień widok
| Rozmiar/SKU | Dostępność | Cena netto |
|---|
Informacje o tej pozycji
NACRES:
NB.72
UNSPSC Code:
41103800
Manufacturer/tradename:
Benchmark Scientific IPS2050-40-E
Informacje o cenach i dostępności nie są obecnie dostępne.
Pomoc techniczna
Pozwól nam pomócpackaging
pack of 1 ea
manufacturer/tradename
Benchmark Scientific IPS2050-40-E
parameter
30-2200 rpm speed
General description
Benchmark′s Overhead Stirrers feature high torque and advanced digital microprocessor control for smooth mixing of low and medium viscosity samples. Speed and run time are accurately set and monitored on the LCD screen. They are supplied with a 4 arm stainless steel propeller, additional steel and PTFE accessories are available. A feed-through chuck holds accessory shafts firmly in place and is easily tightened and loosened without tools. The overhead stirrers are protected from damage by sensors that shut down operation in the case of high viscosity or obstruction, while a splash cover protects the connection mechanism from potential corrosion. Unlike most mixers, the Benchmark series is also supplied with a support stand and mounting hardware. The stirrer easily and securely attaches to the stand for compatibility with a variety of vessels, up to 24 inches high.
Features and Benefits
LCD control panelIncludes complete stand assemblyCompact and quietDigital accuracy and convenience
1 of 1
Ta pozycja | |||
|---|---|---|---|
| description AC/DC input 220 V, Schuko plug | description 4-arm, stainless steel | description flat blade, stainless steel | description AC/DC input 220 V, Schuko plug |
| manufacturer/tradename Benchmark Scientific IPS2050-40-E | manufacturer/tradename Benchmark Scientific IPS2050-P-S4 | manufacturer/tradename Benchmark Scientific IPS2050-P-S1 | manufacturer/tradename Benchmark Scientific IPS2050-20-E |
| packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea | packaging pack of 1 ea |
| parameter 30-2200 rpm speed | parameter - | parameter - | parameter 30-2200 rpm speed |
Still not finding the right product?
Explore all of our products under Benchmark OS-40L Overhead Stirrer
Wybierz jedną z najnowszych wersji:
Masz już ten produkt?